Skip to Content
Merck
All Photos(2)

Key Documents

SAB2500216

Sigma-Aldrich

Anti-CD274/PD-L1 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-B7-H, Anti-B7H1, Anti-PD-L1, Anti-PDCD1LG1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

flow cytometry: suitable
immunofluorescence: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD274(29126)

General description

Programmed cell death 1 ligand 1 (PD-L1), which is also known as cluster of differentiation (CD274) or B7 homolog 1 (B7-H1), is a member of the growing B7 family of immune molecules. PD-L1 is a cell surface immunoglobulin with two Ig-like domains within the extracellular region and a short cytoplasmic domain. The protein is highly expressed in the heart, skeletal muscle, placenta and lung and weakly expressed in the thymus, spleen, kidney and liver. PD-L1 is expressed on activated T-cells, B-cells, dendritic cells, keratinocytes and monocytes. The gene encoding it is localized on human chromosome 9p24.1.

Immunogen

Peptide with sequence CKKQSDTHLEET from the C Terminus of the protein sequence according to NP_054862.1.

Biochem/physiol Actions

Programmed cell death 1 ligand 1 (PD-L1) is involved in the regulation of cellular and humoral immune responses. PD-L1 is up-regulated on T- and B-cells, dendritic cells, keratinocytes and monocytes after lipopolysaccharide (LPS) and interferon-γ (IFNG) activation and up-regulated in B-cells activated by surface Ig cross-linking. PD-L1 is involved in the co-stimulatory signal, essential for T-cell proliferation and production of interleukin-10 (IL10) and IFNG, in an interleukin-2-dependent and a programmed cell death 1 (PDCD1)-independent manner.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Integrative analysis reveals selective 9p24.1 amplification, increased PD-1 ligand expression, and further induction via JAK2 in nodular sclerosing Hodgkin lymphoma and primary mediastinal large B-cell lymphoma.
Green MR
Blood, 116(17), 3268-3277 (2010)
The role of B7 family molecules in hematologic malignancy.
Greaves P and Gribben JG
Blood, 121(5), 734-744 (2013)
Intrinsic and extrinsic control of expression of the immunoregulatory molecule PD-L1 in epithelial cells and squamous cell carcinoma.
Ritprajak P and Azuma M
Oral Oncology, 51(3), 221-228 (2015)
Adnan R Khan et al.
Nature communications, 6, 5997-5997 (2015-01-23)
Specific B-cell subsets can regulate T-cell immune responses, and are termed regulatory B cells (Breg). The majority of Breg cells described in mouse and man have been identified by IL-10 production and are known to suppress allergy and autoimmunity. However

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service