Skip to Content
Merck
All Photos(1)

Key Documents

R6267

Sigma-Aldrich

RANTES human

≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

Regulated on Activation, Normal T-cell Expressed and Secreted

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

≥97% (SDS-PAGE)

form

lyophilized powder

potency

1-5 ng/mL EC50

mol wt

7.8-8.7 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... CCL5(6352)

Looking for similar products? Visit Product Comparison Guide

General description

Regulated on activation, normal T cell expressed and secreted (RANTES), also known as C-C motif chemokine ligand 5 (CCL5), is encoded by the gene mapped to human chromosome 17. RANTES belongs to the chemokine family of proinflammatory basic polypeptides and is expressed by various hematopoietic and non-hematopoietic cell types involved in homing and migration of effector and memory T cells during acute infections.

Biochem/physiol Actions

Regulated on activation, normal T cell expressed and secreted (RANTES) is a chemoattractant towards monocytes, dendritic cells (DCs), memory T cells (CD4+/CD45RO), basophils, and eosinophils. The encoded chemokine protein functions as a ligand for CCR1, CCR3, and CCR5. RANTES inhibits the action of R5 strains of HIV-1 via blocking CCR5 receptor. A variant of RANTES gene is linked with susceptibility to HIV-1 infection and disease progression. Hence, this can be used as a potent biomarker to determine the prognosis of HIV -1 infection.

Physical form

Lyophilized from a 0.2 μm filtered solution in 35% acetonitrile and 0.1% trifluoroacetic acid containing 0.5 mg bovine serum albumin.

Analysis Note

The biological activity is tested in culture using Baf/3 cells. Chemotaxis: EC50: 1-5 ng/mL

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frequency of diagnosis of malignant melanoma.
Roth-Moyo L
Canadian Medical Association Journal (1977)
Role of chemokine and cytokine polymorphisms in the progression of HIV-1 disease.
Biochemical and Biophysical Research Communications (2010)
A role for the chemokine RANTES in regulating CD8 T cell responses during chronic viral infection.
Crawford A
PLoS Pathogens (2011)
RANTES-activated human T lymphocytes. A role for phosphoinositide 3-kinase.
Journal of Immunology (1995)
T J Schall et al.
Nature, 347(6294), 669-671 (1990-10-18)
An important process in the immune response is the migration of different populations of lymphocytes at the proper time to sites of antigenic challenge. Although several chemoattractants are known for broad classes of lymphocytes, such as T and B cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service