Skip to Content
Merck
All Photos(1)

Documents

P1189

Sigma-Aldrich

Polymyxin B

liquid, suitable for cell culture, BioReagent

Synonym(s):

Aerosporin, PMB, PMX, Polymyxin, Polymyxin B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.76

product name

Polymyxin B ready made solution, Cell culture tested, 17 mg/mL in H2O

Quality Level

form

liquid

concentration

17 mg/mL in H2O

storage temp.

2-8°C

General description

Polymyxin B (PMB) is a cyclic cationic lipopeptide antibiotic originally produced by soil bacterium Bacillus polymyxa. Polymyxin B binds specifically to gram negative bacteria by interacting with the anionic lipopolysaccharide (LPS, or endotoxin). Polymyxin B interferes with the outer membrane normal function causing leakage and enhanced uptake of the antibiotic.

Polymyxin B has several applications:
  • It is used to neutralize LPS contamination in vitro and in vivo. Its neutralizing effect is dose-dependent. In vitro experiments showed that PMB inhibits a septic shock by endotoxin inactivation.
  • Cell cultures can be treated with PMB (100U/mL) to avoid early contaminations. PMB is considered a broad spectrum antibiotic and is used to treat infections caused by bacteria resistant to a combination of Penicillin and Streptomycin.
  • PMB is also used as negative and positive control in minimum microbicidal concentration (MIC) assay.

Working concentration:
The suggested concentration of PMB in cell culture mediums is 100U/mL. Therefore, it is recommended to dilute PMB ready made solution 1:1000.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service