Skip to Content
Merck
All Photos(3)

Key Documents

O9139

Sigma-Aldrich

N-(3-Oxododecanoyl)-L-homoserine lactone

≥98% (TLC)

Synonym(s):

3-Oxo-C12-HSL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H27NO4
CAS Number:
Molecular Weight:
297.39
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Product Name

N-(3-Oxododecanoyl)-L-homoserine lactone, quorum sensing signaling molecule

Assay

≥98% (TLC)

form

powder

color

white

application(s)

cell analysis

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCCCCCCCCC(=O)CC(=O)N[C@H]1CCOC1=O

InChI

1S/C16H27NO4/c1-2-3-4-5-6-7-8-9-13(18)12-15(19)17-14-10-11-21-16(14)20/h14H,2-12H2,1H3,(H,17,19)/t14-/m0/s1

InChI key

PHSRRHGYXQCRPU-AWEZNQCLSA-N

Application

N-(3-Oxododecanoyl)-L-homoserine lactone (3-Oxo-C12-HSL), a member of a family of acyl homoserine lactones, may be used to determine its mechanisms and specificity of action as a signaling molecule involved in the regulation of bacterial quorum sensing.

Biochem/physiol Actions

N-(3-oxododecanoyl)homoserine-L-lactone (3-oxo-C12-HSL) is among a group of homoserine lactones that includes: N-octanoyl-homoserine lactone (N-C8-HSL), N-(3-oxodecanoyl) homoserine-L-lactone (3-oxo-C10 HSL), N-(3-Oxotetradecanoyl)-L-homoserine lactone (3-oxo-C14-HSL, N-(3-hydroxydecanoyl)-L-homoserine lactone, and N-(3-hydroxyoctanoyl)-L-homoserine lactone involved in the processes of bacterial quorum sensing. These N-acyl-homoserine lactones are used to study the processes and mechanisms of bacterial quorum sensing.
Acyl homoserine lactone that is an autoinducer of quorum sensing by Pseudomonas aeruginosa. Degraded by paraoxonase (PON) family members.

also commonly purchased with this product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephanie J Cole et al.
Nature communications, 9(1), 4436-4436 (2018-10-27)
Chronic bacterial infections on medical devices, including catheter-associated urinary tract infections (CAUTI), are associated with bacterial biofilm communities that are refractory to antibiotic therapy and resistant to host immunity. Previously, we have shown that Pseudomonas aeruginosa can cause CAUTI by
Gene PA2449 Is Essential for Glycine Metabolism and Pyocyanin
Biosynthesis in Pseudomonas aeruginosa PAO1
Lundgren BR, et al.
Journal of Bacteriology, 195(9), 2087-2100 (2013)
Robust multicellular computing using genetically encoded NOR gates and chemical `wires?
Tamsir A, et al.
Nature, 469(7329), 212-215 (2011)
Megha Shah et al.
Molecular cell, 81(3), 571-583 (2021-01-08)
The arms race between bacteria and phages has led to the evolution of diverse anti-phage defenses, several of which are controlled by quorum-sensing pathways. In this work, we characterize a quorum-sensing anti-activator protein, Aqs1, found in Pseudomonas phage DMS3. We
Kaihao Tang et al.
Scientific reports, 3, 2935-2935 (2013-10-15)
Quorum sensing (QS) is a population-dependent mechanism for bacteria to synchronize social behaviors such as secretion of virulence factors. The enzymatic interruption of QS, termed quorum quenching (QQ), has been suggested as a promising alternative anti-virulence approach. In order to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service