Skip to Content
Merck
All Photos(1)

Key Documents

I8638

Sigma-Aldrich

Anti-Integrin β1 antibody, Mouse monoclonal

clone W1B10, purified from hybridoma cell culture

Synonym(s):

Anti-CD29

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified from hybridoma cell culture
purified immunoglobulin

antibody product type

primary antibodies

clone

W1B10, monoclonal

form

buffered aqueous solution

mol wt

antigen 100-140 kDa

contains

15 mM sodium azide

species reactivity

chicken

technique(s)

immunoprecipitation (IP): suitable
indirect immunofluorescence: suitable
western blot: 1:200 using denatured, non-reduced extract of cultured fibroblasts from chicken embryo lower leg

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

chicken ... ITGB1(374058)

General description

Monoclonal anti-β-1-Chicken Integrin (mouse IgG1 isotype) is derived from the W1B10 hybridoma1 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with purified chicken integrin from fibroblasts of 11 days old chicken embryos. Chicken integrin is a heterodimer consisting of the common β-1 subunit and two α subunits (α-5 or α-3). The β-1 subunit of chicken integrin is a 803-amino acid transmembrane glycoprotein consisting of a large extracellular domain and a short cytoplasmic domain.

Specificity

The antibody recognizes an epitope on the extracellular side of the avian integrin complex, and inhibits adhesion of embryo and tendon fibroblasts as well as skeletal myoblasts to both laminin and fibronectin. Under reducing and denaturing conditions using extracts of chicken embryo muscle cells, the antibody precipitates the β1 class integrin heterodimer as a broad band of ~140 kDa. In the absence of reducing agents, the integrin β1 subunit appears as bands at 100 kDa (immature form), 100 kDa (mature form), and 140 kDa (noncovalently associated α-subunits). The antibody may also be used for bioactivity studies in adhesion assays.

Immunogen

chicken integrin.1

Application

Anti-Integrin β1 antibody, Mouse monoclonal has been used in western blot, fluorescent activated cell sorter (FACS) analysis.

Biochem/physiol Actions

Integrin binding to extracellular matrix (ECM) ligands regulates cell adhesion, shape, motility, intracellular signalling and gene expression. The monoclonal anti-b-1 chicken integrin antibody (clone W1B10) has been used to inhibit chicken muscle cell adhesion in cell cultures.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 1% bovine serum albumin with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bonnie M Marsick et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 32(1), 282-296 (2012-01-06)
The development of a functioning neural network relies on responses of axonal growth cones to molecular guidance cues that are encountered en route to their target tissue. Nerve growth factor (NGF) and neurotrophin-3 serve as attractive cues for chick embryo
beta1-Integrin-collagen interaction reduces chondrocyte apoptosis
Cao L, et al.
Matrix Biology, 18(4), 343-355 (1999)
Sarah M Short et al.
The Journal of cell biology, 168(4), 643-653 (2005-02-18)
The anti-angiogenic effect of thrombospondin-1 has been shown to be mediated through binding of the type-1 repeat (TSR) domain to the CD36 transmembrane receptor. We now report that the TSR domain can inhibit VEGF-induced migration in human umbilical vein endothelial
Masayuki Yamashita
Communications biology, 6(1), 680-680 (2023-07-01)
Retinal ganglion cell (RGC) axons converge on the optic disc to form an optic nerve. However, the mechanism of RGC axon convergence remains elusive. In the embryonic retina, an electric field (EF) exists and this EF converges on the future
Leticia Labriola et al.
BMC cell biology, 10, 49-49 (2009-06-24)
The in vitro culture of insulinomas provides an attractive tool to study cell proliferation and insulin synthesis and secretion. However, only a few human beta cell lines have been described, with long-term passage resulting in loss of insulin secretion. Therefore

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service