Skip to Content
Merck
All Photos(1)

Key Documents

I8026

Sigma-Aldrich

Anti-Interleukin-8 antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-CXCL8, Anti-IL-8

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

immunocytochemistry: 5-15 μg/mL
immunohistochemistry: 5-15 μg/mL
neutralization: suitable
western blot: 0.1 μg/mL

storage temp.

−20°C

Gene Information

human ... IL8(3576)

Looking for similar products? Visit Product Comparison Guide

General description

Interleukin 8 (IL-8) is an 8kD, 72 amino acid residue containing cytokine that belongs to C-X-C chemokine family . The IL-8 gene is mapped to human chromosome 4q13.3.

Specificity

The antibody will neutralize the biological activity of recombinant and natural human IL-8, but not recombinant human GROα.

Immunogen

recombinant human IL-8 (CXCL8).

Application

Anti-Interleukin-8 antibody can be used for the following :
  • For neutralizing the biological activity of recombinant human IL-8 and natural human IL-8.
  • Immunoblotting.
  • ELISA.
Anti-Interleukin-8 antibody can be used in immunocytochemistry and immunohistochemistry.

Biochem/physiol Actions

Interleukin 8 (IL-8) stimulates neutrophils to produce lysosomal enzymes along with myeloperoxidase, α-mannosidase and β-glucuronidase . Goat anti-Interleukin-8 antibody reacts specifically with recombinant and natural human IL-8 but not with recombinant human GROα. Elevated expression of the gene is observed in a number of cancers including chronic spontaneous urticaria and breast cancer, thus serves as a prognostic marker for the same. IL-8 regulates the metastasis and angiogenesis processes during cancer development. IL-8 is responsible for epithelial–mesenchymal transition which plays an important role in providing cancer cells the mobility to invade other neighbouring cells, thereby contributing to metastasis.

Physical form

Lyophilized from 0.2 μm filtered solution in phosphate buffered saline containing carbohydrates.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification and validation of vesicant therapeutic targets using a high-throughput siRNA screening approach.
Ruff A L, et al.
Archives of Toxicology, 90(2), 375-383 (2016)
Elevated plasma il-8 concentration is related to severity of systemic inflammation in chronic spontaneous urticaria.
Kasperska-Zajac A, et al.
Journal of Biological Regulators and Homeostatic Agents, 31(4), 957-961 (2017)
Properties of pro-inflammatory cell type-specific leukocyte chemotactic cytokines, interleukin 8 (IL-8) and monocyte chemotactic and activating factor (MCAF).
N Mukaida et al.
Microbiology and immunology, 36(8), 773-789 (1992-01-01)
T Yoshimura et al.
Proceedings of the National Academy of Sciences of the United States of America, 84(24), 9233-9237 (1987-12-01)
Stimulated human monocytes release several proteins thought to play a role in inflammation, including interleukin 1, tumor necrosis factor, and plasminogen activator. We have purified another proinflammatory protein that is chemotactic for human neutrophils from conditioned medium of lipopolysaccharide-stimulated monocytes.
Sohlh2 suppresses epithelial to mesenchymal transition in breast cancer via downregulation of IL-8.
Ji S, et al.
Oncotarget, 7(31), 49411-49411 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service