Skip to Content
Merck
All Photos(6)

Key Documents

HPA038109

Sigma-Aldrich

Anti-FAM13A antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FAM13A1, Anti-Family with sequence similarity 13, member A, Anti-KIAA0914

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

TDFSARCFLDQFEDDADGFISPMDDKIPSKCSQDTGLSNLHAASIPELLEHLQEMREEKKRIRKKLRDFE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FAM13A(10144)

General description

Family with sequence similarity 13 member A (FAM13A), also known as FAM13A1, is encoded by the gene mapped to human chromosome 4q22.1. The encoded protein contains Rho GTPase-activating protein (Rho-GAP) domain.

Immunogen

family with sequence similarity 13, member A recombinant protein epitope signature tag (PrEST)

Application

Anti-FAM13A antibody produced in rabbit has been used in immunohistochemistry and western blot analysis.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

Family with sequence similarity 13 member A (FAM13A) plays a vital role in signal transduction. Mutation in the gene is linked with chronic obstructive pulmonary disease (COPD). The encoded protein can regulate the stability of β-catenin. In addition, it also regulates growth and progression of non-small cell lung cancer (NSCLC).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST79751

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Variants in FAM13A are associated with chronic obstructive pulmonary disease.
Cho MH
Nature Genetics, 42, 200-202 (2010)
A Chronic Obstructive Pulmonary Disease Susceptibility Gene, FAM13A, Regulates Protein Stability of ?-Catenin.
Jiang Z
American Journal of Respiratory and Critical Care Medicine, 194, 185-197 (2016)
Jinyuan Zhu et al.
Respiratory research, 22(1), 192-192 (2021-07-03)
To explore the role of family with sequence similarity 13 member A (FAM13A) in TGF-β1-induced EMT in the small airway epithelium of patients with chronic obstructive pulmonary disease (COPD). Small airway wall thickness and protein levels of airway remodeling markers
FAM13A is associated with non-small cell lung cancer (NSCLC) progression and controls tumor cell proliferation and survival.
Eisenhut F
Oncoimmunology, 6 (2016)
FAM13A locus in COPD is independently associated with lung cancer - evidence of a molecular genetic link between COPD and lung cancer.
Young RP
The Application of Clinical Genetics, 4, 1-10 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service