Skip to Content
Merck
All Photos(5)

Documents

HPA035866

Sigma-Aldrich

Anti-RBP2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CRABP-II, Anti-CRBP2, Anti-CRBPII, Anti-RBPC2, Anti-Retinol binding protein 2, cellular

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

RKIAVRLTQTKVIDQDGDNFKTKTTSTFRNYDVDFTVGVEFDEYTKSLDNRHVKALVTWEGDVLVC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RBP2(5948)

General description

The gene RBP2 (retinol-binding protein 2) is mapped to human chromosome 3q23. It is mainly expressed in the small intestine and is a cytosolic protein. . It belongs to the CRBP (cellular retinoic-acid-binding protein) intracellular lipid-binding protein family.

Immunogen

retinol binding protein 2, cellular recombinant protein epitope signature tag (PrEST)

Application

Anti-RBP2 antibody produced in rabbit has been used in Western Blotting and immunocytochemistry.

Biochem/physiol Actions

RBP2 (retinol-binding protein 2) is responsible for vitamin A absorption/uptake. It works as a cellular retinoid chaperone. RBP2 associates with retinol and retinal, thereby assisting in retinoid homeostasis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST79303

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cellular retinol-binding protein type II (CRBPII) in adult zebrafish (Danio rerio). cDNA sequence, tissue-specific expression and gene linkage analysis.
Cameron MC
European Journal of Biochemistry, 269(18), 4685-4692 (2002)
Massive bowel resection upregulates the intestinal mRNA expression levels of cellular retinol-binding protein II and apolipoprotein A-IV and alters the intestinal vitamin A status in rats.
Hebiguchi T
International Journal of Molecular Medicine, 35(3), 724-730 (2015)
Crystal structure of human cellular retinol-binding protein II to 1.2 A resolution.
Tarter M
Proteins: Structure, Function, and Genetics, 70(4), 1626-1630 (2008)
Taku Hebiguchi et al.
International journal of molecular medicine, 35(3), 724-730 (2015-01-15)
Short bowel (SB) syndrome causes the malabsorption of various nutrients. Among these, vitamin A is important for a number of physiological activities. Vitamin A is absorbed by epithelial cells of the small intestine and is discharged into the lymphatic vessels
Hepatocyte nuclear factor-4alpha regulates human cellular retinol-binding protein type II gene expression in intestinal cells.
Yamaguchi N
American Journal of Physiology: Gastrointestinal and Liver Physiology, 296(3), G524-G533 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service