Skip to Content
Merck
All Photos(5)

Key Documents

HPA003590

Sigma-Aldrich

Anti-NDRG2 antibody produced in rabbit

enhanced validation

Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Protein NDRG2 antibody produced in rabbit, Anti-Protein Syld709613 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

THSVETPYGSVTFTVYGTPKPKRPAILTYHDVGLNYKSCFQPLFQFEDMQEIIQNFVRVHVDAPGMEEGAPVFPLGYQYPSLDQLADMIPCVLQYLNFSTIIGVGVGAGA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NDRG2(57447)

Looking for similar products? Visit Product Comparison Guide

General description

The NDRG2 (N-Myc downstream-regulated gene 2) gene is mapped to human chromosome 14q11.2. It belongs to the family of N-Myc downstream-regulated genes. The gene contains 16 exons interspaced by 15 introns. It is found to be expressed abundantly in many normal tissues including brain, heart, skeletal muscle, and kidney. Its expression is observed to be very low in cancers and cancer cell lines.

Immunogen

Protein NDRG2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

N-myc downstream-regulated gene 2 is a protein encoded by the NDRG2 gene in humans. It is a new tumor suppressor gene and plays an important role in tumor suppression in several cancers or cancer cell lines. The gene plays a crucial role in controlling tumor growth. It functions as one of the candidate genes in pancreatic carcinogenesis and has physiological roles in neuroblastoma cell proliferation. It may act as a candidate for the treatment of neuroblastoma in children.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70587

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhi-Guo Zhang et al.
Asian Pacific journal of cancer prevention : APJCP, 15(1), 239-244 (2014-02-18)
Several recent studies have showed that the n-myc downstream regulated gene 2 (NDRG2) is a new tumor suppressor gene, and that it plays an important role in tumor suppression in several cancers or cancer cell lines. However, few studies focused
Ruixiao Li et al.
PloS one, 8(10), e76689-e76689 (2013-10-23)
N-Myc downstream-regulated gene 2 (NDRG2) is a candidate tumor suppressor gene, which plays an important role in controlling tumor growth. The aim of this study was to investigate the expression of NDRG2 gene in bladder cancer (BC) tissues and several
Xiao-Lan Hu et al.
World journal of gastroenterology, 10(23), 3518-3521 (2004-11-05)
To investigate the expression of NDRG2 and mutation of the entire coding region of NDRG2 in human liver and pancreatic cancers, and to further discuss the possible causes of NDRG2 distinct expression patterns. Reverse transcription-polymerase chain reaction (RT-PCR) was used
Akihiro Yamamura et al.
Biochemical and biophysical research communications, 441(1), 102-107 (2013-10-19)
Pancreatic cancer is a highly lethal disease with a poor prognosis; the molecular mechanisms of the development of this disease have not yet been fully elucidated. N-myc downstream regulated gene 2 (NDRG2), one of the candidate tumor suppressor genes, is
Ji Ma et al.
Breast cancer research : BCR, 16(2), R27-R27 (2014-03-19)
N-myc downstream-regulated gene 2 (NDRG2), a novel tumour suppressor and cell stress-related gene, is involved in many cell metabolic processes, such as hormone, ion and fluid metabolism. We investigated whether NDRG2 is involved in any glucose-dependent energy metabolism, as well

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service