Skip to Content
Merck
All Photos(7)

Documents

HPA003353

Sigma-Aldrich

Anti-SLC16A2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MCT 7, Anti-MCT 8, Anti-Monocarboxylate transporter 8, Anti-Solute carrier family 16 member 2, Anti-X-linked PEST-containing transporter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:500-1:1000

immunogen sequence

SSRNKYQPQSGSSGPSSHSPPAAMALQSQASEEAKGPWQEADQEQQEPVGSPEPESEPEPEPEPEPVPVPPPEPQPEPQPLPDPAPLPELEFESERVHEPEPT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC16A2(6567)

Immunogen

Monocarboxylate transporter 8 recombinant protein epitope signature tag (PrEST)

Application

Anti-SLC16A2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

SLC16A2 (solute carrier family 16, member 2) gene encodes a carrier protein that specifically transports thyroid hormone. It functions in the cellular uptake of thyroxine (T4), triiodothyronine (T3), reverse triiodothyronine (rT3) and diidothyronine (T2). Mutations in this gene cause Allan-Herndon-Dudley syndrome (AHDS), an X-linked developmental disorder characterized by hypotonia, spasticity, muscle hypoplasia, neurological disorders, cognitive impairment and severe neurological defects. Defects in this gene also cause learning disability and motor or movement disorder in males.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74389

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad Saiful Islam et al.
Endocrine journal, 66(1), 19-29 (2018-10-30)
Monocarboxylate transporter 8 (MCT8) facilitates T3 uptake into cells. Mutations in MCT8 lead to Allan-Herndon-Dudley syndrome (AHDS), which is characterized by severe psychomotor retardation and abnormal thyroid hormone profile. Nine uncharacterized MCT8 mutations in Japanese patients with severe neurocognitive impairment
Hideyuki Iwayama et al.
Thyroid : official journal of the American Thyroid Association, 26(9), 1311-1319 (2016-07-20)
MCT8 gene mutations produce thyroid hormone (TH) deficiency in the brain, causing severe neuropsychomotor abnormalities not correctable by treatment with TH. This proof-of-concept study examined whether transfer of human MCT8 (hMCT8) cDNA using adeno-associated virus 9 (AAV9) could correct the
Lori M Roberts et al.
Endocrinology, 149(12), 6251-6261 (2008-08-09)
Thyroid hormones require transport across cell membranes to carry out their biological functions. The importance of transport for thyroid hormone signaling was highlighted by the discovery that inactivating mutations in the human monocarboxylate transporter-8 (MCT8) (SLC16A2) cause severe psychomotor retardation
Yoshiyuki Henning et al.
Frontiers in cellular neuroscience, 10, 205-205 (2016-09-13)
The thyroid hormones (TH) triiodothyronine (T3) and its prohormone thyroxine (T4) are crucial for retinal development and function, and increasing evidence points at TH dysregulation as a cause for retinal degenerative diseases. Thus, precise regulation of retinal TH supply is
W Edward Visser et al.
Human mutation, 30(1), 29-38 (2008-07-19)
Monocarboxylate transporter 8 (MCT8; approved symbol SLC16A2) facilitates cellular uptake and efflux of 3,3',5-triiodothyronine (T3). Mutations in MCT8 are associated with severe psychomotor retardation, high serum T3 and low 3,3',5'-triiodothyronine (rT3) levels. Here we report three novel MCT8 mutations. Two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service