Skip to Content
Merck
All Photos(1)

Key Documents

H4290

Sigma-Aldrich

Hydroxyphenyl fluorescein solution

5 mM in DMF

Synonym(s):

Hydroxyphenyl fluorescein, 2-[6-(4′-hydroxy)phenoxy-3H-xanthen-3-on-9-yl]benzoic acid, HPF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H16O6
Molecular Weight:
424.40
EC Number:
UNSPSC Code:
12352108
NACRES:
NA.32

concentration

5 mM in DMF

fluorescence

λex 490 nm; λem 515 nm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Hydroxyphenyl fluorescein solution, also known as HPF is a fluorescent probe, which upon oxidation by the hydroxyl radical (HO) returns a brilliant green fluorescence (HPF-ox). Increased HPF signals in antibiotic-exposed bacterial cells are explained by fluorescence associated with increased cell size, not by ROS levels. In cells incubated with the HPF probe and exposed to ampicillin and norfloxacin with HPF fluorescence correlated linearly.

Application

Fluorescent reagent for the detection of highly reactive oxygen species (hROS). Fluorescence intensity increases after the immediate reaction with hROS such as hydroxyl radical and peroxynitrite. Since HPF does not react with nitric oxide, superoxide and hydrogen peroxide, it can differentially detect peroxynitrite. HPF is resistant to light-induced autoxidation.
Hydroxyphenyl fluorescein solution is used to detect highly reactive oxygen species (hROS). It is a suitable probe to detect cellular death caused by bacterial antibiotics.

Features and Benefits

The following are the benefits of Hydroxyphenyl fluorescein solution:

  • HPF is resistant to light-induced autooxidation.
  • Hypochlorite(-OCl) does not react with HPF.
  • HPF does not react with nitric oxide, superoxide, and hydrogen peroxide. 
  • HPF has low reactivity with HOCl.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

135.5 °F - (Dimethylformamide)

Flash Point(C)

57.5 °C - (Dimethylformamide)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ken-ichi Setsukinai et al.
The Journal of biological chemistry, 278(5), 3170-3175 (2002-11-07)
We designed and synthesized 2-[6-(4'-hydroxy)phenoxy-3H-xanthen-3-on-9-yl]benzoic acid (HPF) and 2- [6-(4'-amino)phenoxy-3H-xanthen-3-on-9-yl]benzoic acid (APF) as novel fluorescence probes to detect selectively highly reactive oxygen species (hROS) such as hydroxyl radical (*OH) and reactive intermediates of peroxidase. Although HPF and APF themselves scarcely
Michael A Kohanski et al.
Cell, 130(5), 797-810 (2007-09-07)
Antibiotic mode-of-action classification is based upon drug-target interaction and whether the resultant inhibition of cellular function is lethal to bacteria. Here we show that the three major classes of bactericidal antibiotics, regardless of drug-target interaction, stimulate the production of highly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service