Skip to Content
Merck
All Photos(1)

Documents

GW21184P

Sigma-Aldrich

Anti-c-Myc (ab1) antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Myc proto-oncogene protein

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, mouse, human

manufacturer/tradename

Genway 15-288-21184P

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYC(4609)

Related Categories

Immunogen

Immunogen Sequence: GI # 12962935, sequence 410-419
Synthetic peptide of V-myc myelocytomatosis viral oncogene homolog.

Application

Anti-c-Myc (ab1) antibody produced in chicken is suitable for indirect ELISA and western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Myc (c-Myc) is a regulatory gene and encodes for a transcription factor. It is a multifunctional, nuclear phosphoprotein that plays a role in cell cycle progression, apoptosis and cellular transformation. Myc is associated with aggressive behaviour in several cancers. c-MYC gain with lung adenocarcinoma is a candidate for additional first-line treatment to mitigate increased risk for disease progression and death. It plays an important role in the pathogenesis of B-cell prolymphocytic leukemia (B-PLL).

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ellen Flatley et al.
American journal of clinical pathology, 142(3), 347-354 (2014-08-16)
B-cell prolymphocytic leukemia (B-PLL) remains a controversial entity, and its molecular pathogenesis is largely unknown. Patients are older, typically having marked lymphocytosis and splenomegaly in the absence of lymphadenopathy. It is defined as a mature B-cell leukemia with more than
A N Seo et al.
British journal of cancer, 110(11), 2688-2699 (2014-05-09)
c-MYC copy number gain (c-MYC gain) has been associated with aggressive behaviour in several cancers. However, the role of c-MYC gain has not yet been determined in lung adenocarcinomas classified by genetic alterations in epidermal growth factor receptor (EGFR), KRAS
Enrico Alessio et al.
Nucleic acids research, 47(4), 1653-1670 (2019-01-17)
Long non-coding RNAs (lncRNAs) are emerging as important players in the regulation of several aspects of cellular biology. For a better comprehension of their function, it is fundamental to determine their tissue or cell specificity and to identify their subcellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service