Skip to Content
Merck
All Photos(1)

Key Documents

GE10600022

Amersham Hybond® LFP Western blotting membranes, PVDF

pore size 0.2 μm, roll W × L 254 mm × 4 m, pkg of 1 ea

Synonym(s):

PVDF membranes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105300
NACRES:
NB.22

packaging

pkg of 1 ea

manufacturer/tradename

Cytiva 10600022

roll W × L

254 mm × 4 m

pore size

0.2 μm pore size

Looking for similar products? Visit Product Comparison Guide

General description

Low-fluorescent, hydrophobic PVDF protein blotting membrane with 0.2 µm pore size. Amersham Hybond LFP 0.2 PVDF is a low-fluorescent, hydrophobic membrane, with 0.2 µm pore size, for protein transfer. Amersham Hybond LFP is optimized for fluorescent detection in Western blotting applications and for use with the Amersham ECL Plex detection system. This membrane has low background fluorescence, resulting in high sensitivity. Typical protein binding capacity is > 200 µg/cm2. The PVDF protein blotting membrane, with 0.2 µm pore size, provides low auto-fluorescence across a wide range of excitation/emission wavelengths.

Features and Benefits

  • Much lower background fluorescence than other commercially available PVDF membranes, resulting in higher sensitivity.
  • Protein binding capacity over 200 μg/cm2.
  • Compatible with a variety of blocking agents, transfer buffers, and total protein blot stains.
  • Available in popular precut sizes for laboratory applications.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Legal Information

Amersham is a trademark of Cytiva
Hybond is a registered trademark of Cytiva

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raúl Valdes et al.
Neurotoxicity research, 39(2), 182-190 (2021-02-09)
The enzyme glutathione transferase M2-2, expressed in human astrocytes, increases its expression in the presence of aminochrome and catalyzes the conjugation of aminochrome, preventing its toxic effects. Secretion of the enzyme glutathione transferase M2-2 from U373MG cells, used as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service