EP5305000703
Eppendorf® Concentrator Plus
complete system, integrated vacuum pump, external device junction, without rotor, AC/DC input 230 V, EuroPlug
Synonym(s):
Centrifuge
About This Item
Recommended Products
feature
CE compliant
CSA approved
to hold 144 × 0.5 mL tubes OR 96 × 1.5/2 mL OR 6 × 50 mL conical tubes OR 2 × microplate/PCR plates
complete system, integrated vacuum pump, external device junction, without rotor
manufacturer/tradename
Eppendorf® 5305000703 (EU)
parameter
ambient
1400 rpm speed
30 °C temp. range (45 °C) at 60 °C
W × D × H
33 cm × 58 cm × 29 cm
weight
31.5 kg
Looking for similar products? Visit Product Comparison Guide
General description
- The basic Concentrator plus system for connection to an external vacuum pump
- The Concentrator plus complete system with an integrated, solvent-resistant, maintenance, and oil-free PTFE diaphragm vacuum pump and condensation trap
- The same Concentrator plus complete system, but with an extra vacuum port added so you can use the integrated pump separately without having to disconnect it
Application
Features and Benefits
- Extremely quiet operation <50 dB(A)
- Small footprint saves valuable bench space
- Wide variety of 15 different rotor options to accommodate tubes from 0.2 mL to 50 mL, microplates and PCR plates
- Motorless induction drive and chemical-resistant, stainless steel chamber for trouble-free operation
- Can be connected with most commercially available cold traps for use with vapors
- Coated lid with superior chemical resistance
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service