EP0030073851
Eppendorf® epT.I.P.S.® G Reloads, PCR clean
volume (50-1000 μL), L (71 mm), blue, pack of 960 ea (10 trays × 96 tips)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
blue (Tray)
sterility
non-sterile
packaging
pack of 960 ea (10 trays × 96 tips)
manufacturer/tradename
Eppendorf® 0030073851
technique(s)
PCR: suitable
L
(71 mm)
maximum volume
1 mL
volume
(50-1000 μL)
color
blue
Looking for similar products? Visit Product Comparison Guide
General description
The perfect fit for your Eppendorf pipette: epT.I.P.S. ensure a pipetting system you can rely on. Make everyday work easier and more enjoyable with the epT.I.P.S. (Eppendorf Totally Integrated Pipetting System) pipette tips. These pipette tips are manufactured to extremely high quality and purity standards and are designed to cover all of the specific needs of your samples. Offering reduced-force tip attachment and ejection with complete sealing, epT.I.P.S. work in perfect combination with your Eppendorf pipettes and form a fully integrated pipetting system that delivers maximum precision, accuracy and reproducibility. Eppendorf pipette tips with ideal wetting properties and optimum transparency are manufactured exclusively from high-purity plastic materials. The universal nose cone design allows Eppendorf tips to also be used with pipettes from other manufacturers. Pipette tip reloads for insertion into the epT.I.P.S. Box 2.0.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service