Skip to Content
Merck
All Photos(2)

Key Documents

CLS352340

Corning® Falcon® Cell Strainer For Use With 50ml Conical Tubes

pore size 40 μm, blue Nylon, sterile

Synonym(s):

culture tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104932
NACRES:
NB.13
Pricing and availability is not currently available.

material

blue Nylon

sterility

sterile

manufacturer/tradename

Corning 352340

pore size

40 μm pore size

application(s)

filtration

Looking for similar products? Visit Product Comparison Guide

Legal Information

Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service