CLS11405
Corning® Erlenmeyer flask, with aseptic connector
volume 125 mL, I.D. 1/8 in., sterile
Synonym(s):
Glass Erlenmeyer flask
All Photos(1)
About This Item
UNSPSC Code:
41122104
NACRES:
NB.43
Recommended Products
material
flat bottom flask
polycarbonate
polypropylene cap
description
plain
sterility
sterile
feature
cap: no
C-Flex tubing
packaging
case of 6
manufacturer/tradename
Corning 11405
I.D.
1/8 in.
capacity
125 mL
volume
125 mL
fitting
male luer lock
Related Categories
Features and Benefits
- An aseptic transfer cap is preassembled on an optically clear polycarbonate flask
- Graduated for better accuracy
- Cap is made of flexible tubing that is heat-sealable and chemically resistant with male luer lok disconnect
- Gamma-radiation sterilized
- When adding or removing medium and cells, this flask enhances the productivity of the shaker culture operation
- Nonpyrogenic
Other Notes
Browse all Erlenmeyer flasks
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)