Skip to Content
Merck
All Photos(1)

Documents

CLLS1007

Sigma-Aldrich

SW48 Cells TP53 -/-

Human female colon, colorectal adenocarcinoma

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

product name

SW48 Cells TP53 -/-,

biological source

human female colon (Source Disease: Colorectal adenocarcinoma)

Quality Level

OMIM accession no.

storage temp.

−196°C

Gene Information

human ... TP53(7157)

General description

SW48 Cells TP53 -/- are colorectal adenocarcinoma, epithelial cells, from a human caucasian female (aged 82 years), with a ZFN knock out modification.

Application

Media Renewal changes one to two times per week
Rapidly thaw vial by gentle agitation in 37°C water bath (~2 minutes), keeping vial cap out of the water. Decontaminate with 70% ethanol, add 9 mL culture media and centrifuge 125 x g (5-7 minutes). Resuspend in complete culture media and incubate at 37°C in a 5% CO2 atmosphere.
Subculture Ratio: approx. 1:2 to 1:6
The base medium for this cell line is DMEM, Cat. No. D5671. To make the complete growth medium, add the following components to the base medium: fetal bovine serum, Cat. No. F4135, to a final concentration (v/v) of 10%, Sodium pyruvate, Cat. No. S8636, at a final concentration of 1 mM, and L-glutamine, Cat. No. G7513, at a final concentration of 2mM.
The cell freezing medium is DMSO 1X (Cat. No. C6164)

Features and Benefits

These SW48 cells are adherent, with a doubling time of approx. 35 hours.
Zinc finger nuclease (ZFN) knock out on chromosome 17p13.1

Quality

Tested for Mycoplasma, sterility, post-freeze viability, short terminal repeat (STR) analysis for cell line identification, PCR assay for cell line species confirmation.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Kit Components Also Available Separately

Product No.
Description
SDS

  • SW48 Cells TP53 -/- 1 vial

  • SW48 Control Cells 1 vial


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service