Skip to Content
Merck
All Photos(1)

Key Documents

HL60 Cell Line human

98070106, human blood, Lymphoblast

Synonym(s):

HL60 Cell Culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

HL60 Cell Line human, Caucasian promyelocytic leukemia, 98070106

biological source

human blood

growth mode

Suspension

karyotype

Modal no. 46, pseudodiploid

morphology

Lymphoblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian promyelocytic leukaemia

Cell Line Description

The HL-60 cell line was derived from peripheral blood leukocytes obtained by leukopheresis of a 36-year-old Caucasian female with acute promyelocytic leukemia. It was among the first long-term suspension cultures of human myeloid leukaemic cells to be established. Approximately 10% of HL-60 cells spontaneously differentiate and differentiation can be stimulated by polar planar compounds butyrate, hypoxanthine, phorbol myristic acid (PMA, TPA), dimethylsulfoxide (DMSO, 1% to 1.5%), actinomycin D, and retinoic acid.

Application

Differentiation studies
HL60 cell line has been used to study the effect of inecalcitol on the expression of cluster of differentiation 38 (CD38). It has also been used to study the role of autophagy in the cytotoxicity of cytarabine (an anti-leukemic drug).

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 13,14
D13S317: 8,11
D16S539: 11
D5S818: 12
D7S820: 11,12
THO1: 8
TPOX: 8,11
vWA: 16

Culture Medium

RPMI 1640 + 2mM Glutamine + 10-20% Foetal Bovine Serum (FBS).

Subculture Routine

When starting from a frozen ampoule, add thawed cells to a conical based centrifuge tube e.g. 15ml tube. Slowly add 4 ml of culture medium to the tube. Take a sample of the cell suspension, e.g. 100ul, to count cells. Centrifuge the cell suspension at low speed i.e. 100 - 150 x g for a maximum of 5 mintues. Remove medium and resuspend the cell pellet at a density of 3 - 5 x 100,000 cells/ml in fresh medium containing 20% serum. Incubate flask at 37 C; 5% CO2. Cell growth after resuscitation is slow, it may take up to 10 days for proliferation to be established. Check daily. Once the culture is established the serum concentration can be reduced to 10%. Maintain cultures between 1-9 x 100,000 cells/ml; 5% CO2; 37 C; at low density or they may differentiate. At ECACC it has been found to be difficult to recover cells of acceptable viability after freezing in 10% glycerol/90% FBS. We recommend using 10% DMSO/90% FBS for this purpose, but spinning out the cells at resuscitation as above to remove the DMSO as it can cause cells to differentiate. After 6 weeks in culture cells may differentiate.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DNA, RNA, cDNA derived from ECACC mammalian cell lines allow screening for genes or expression patterns to identify lines most suitable for specific research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service