Skip to Content
Merck
All Photos(1)

Documents

COR-L23/5010

96042338, human lung, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

COR-L23/5010, 96042338

biological source

human lung

growth mode

Aggregates in suspension

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian lung, large cell carcinoma, drug resistant

Cell Line Description

COR-L23/5010 has been derived from the parent line, COR-L23 (Sigma Catalogue number. 92031919) by continuous exposure to increasing concentrations of doxorubicin (also known as adriamycin). The cells overexpress multidrug resistance-associated protein (MRP) and express a 190kDa membrane-protein connected to the degree of drug-resistance. COR-L23/5010 have a higher resistance than COR-L23/R (Sigma Catalogue number. 96042339).

Application

MRP studies, drug-resistance studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11
D13S317: 10,11
D16S539: 11,13
D5S818: 10,12
D7S820: 9
THO1: 9
TPOX: 8,11
vWA: 19

Culture Medium

RPMI 1640 + 2mM Glutamine + 1.0μg/ml doxorubicin + 10% FBS

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C It is recommended to culture the cells without drug after resuscitation until the first passage.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service