Skip to Content
Merck
All Photos(1)

Key Documents

Neopu

90070561, insect embryo, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

Neopu, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 90070561

biological source

insect embryo

packaging

tube of 5 μg 90070561-DNA-5UG
pkg of vial of cells 90070561-1VL

growth mode

Semi-adherent

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Drosophila embryo

Cell Line Description

Supports the growth of rhabdovirus and picornavirus. Developed from the embryonic cells of Drosophila melanogaster.

Application

Virus studies: rhabdovirus, picornavirus

Culture Medium

Schneider′s Drosophila medium + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml. Cells may semi-adhere to the tissue culture flask, shake gently to dislodge; 25°C; No CO2.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service