Skip to Content
Merck
All Photos(1)

Key Documents

C-12521

Sigma-Aldrich

Human Pulmonary Artery Smooth Muscle Cells (HPASMC)

500,000 cryopreserved cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

biological source

human pulmonary artery

packaging

pkg of 500,000 cells

morphology

(smooth muscle)

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Cell Line Origin

Artery

Application

Primary Human Pulmonary Artery Smooth Muscle Cells (HPASMC) are isolated from human pulmonary arteries and are provided in a cryopreserved format. HPASMC stain positive for smooth muscle α-actin. They are suited for studying the function of pulmonary arteries under normal and disease conditions (e.g. vasoconstriction induced by alveolar hypoxia). They are also useful for physiological studies of vascular smooth muscle cells.

Quality

Rigid quality control tests are performed for each lot of Human Smooth Muscle Cells. They are tested for cell morphology, adherence rate, and cell viability. Flow cytometric analyses for cell-type specific markers, e.g. smooth muscle a-actin, are carried out for each lot. Growth performance is tested through multiple passages up to 15 population doublings (PD) under culture conditions without antibiotics and antimycotics. In addition, all cells have been tested for the absence of HIV-1, HIV-2, HBV, HCV, HTLV-1, HTLV-2 and microbial contaminants (fungi, bacteria, and mycoplasma).

Warning

Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.

Subculture Routine

Click here for more information.

Other Notes

Recommended Plating Density: 7500 - 10000 cells per cm2Passage After Thawing: P2Tested Markers: Smooth muscle alpha-actin positiveGuaranteed population doublings: >15

Recommended products

Recommended Primary Cell Culture Media:Link

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong-Ling Hu et al.
American journal of respiratory cell and molecular biology, 42(6), 661-666 (2009-07-21)
Chronic hypoxia induces proliferation of human pulmonary artery smooth muscle cells (hPASMCs), leading to remodeling and pulmonary hypertension, but the mechanism remains unclear. The present study tested the roles of mitochondrial ATP-sensitive potassium channel (mitoK(ATP)) and mitochondrial membrane potential (DeltaPsi(m))

Articles

Smooth muscle human primary cells isolated from blood vessels and hollow organs, plus smooth muscle cell growth media. Protocols on media handling, cell passaging and product use.

Smooth muscle human primary cells isolated from blood vessels and hollow organs, plus smooth muscle cell growth media. Protocols on media handling, cell passaging and product use.

Smooth muscle human primary cells isolated from blood vessels and hollow organs, plus smooth muscle cell growth media. Protocols on media handling, cell passaging and product use.

Smooth muscle human primary cells isolated from blood vessels and hollow organs, plus smooth muscle cell growth media. Protocols on media handling, cell passaging and product use.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service