Skip to Content
Merck
All Photos(1)

Key Documents

C1188

Sigma-Aldrich

Bovine Collagen Type II

from bovine tracheal cartilage, powder

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

Product Name

Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder

biological source

bovine cartilage (tracheal)

Quality Level

form

powder

solubility

aqueous acid: soluble

UniProt accession no.

storage temp.

2-8°C

Gene Information

General description

Collagen is classified into a number of structurally and genetically distinct types. We use the nomenclature proposed by Bornstein and Traub. Do not confuse Sigma type designations with recognized collagen classification types.
Collagen protein is abundantly present in human. It is the prime structural constituent of dermis. It is also an important constituent of extracellular matrix (ECM).

Application

Collagen from bovine tracheal cartilage has been used to induce arthritis in male DBA/1 mice. It has also been used as a fluorophore to acquire fluorescence emission spectra on the reflectance and fluorescence lifetime spectrometer (RFLS).
Collagen-type II has been investigated for its importance in haemostasis. Two peptides, CB8 and CB11, are sites of platelet binding and activation on collagen-type II. Collagen-type II has also been used as a model for arthritis in animals, where FR140423, a selective cyclooxygenase (COX)-2 inhibitor, has shown an anti-inflammatory effect without gastric lesions that may occur with treatment via indomethacin.

Collagen Type II is sometimes used to induce an animal model of rheumatoid arthritis. However, under conditions leading to formation of reactive oxygen species, the subjects develop tolerance for auto-antibodies. This is an unusual situation of ROS having a favorable effect.

Biochem/physiol Actions

Collagen participates in the aging process. It offers a structural framework for cells and mechanical strength to tissues.

Preparation Note

Prepared by a modification of the pepsin extraction procedure of Trentham, et al., J. Exp. Med., 146, 857 (1977).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular and cellular basis of hypertrophic scarring
Total Burn Care, 495-505 (2012)
Important role of interleukin-3 in the early phase of collagen-induced arthritis
Bruhl H, et al.
Arthritis and Rheumatism, 60(5), 1352-1361 (2009)
Rosalia Cuahtecontzi Delint et al.
Biomaterials, 276, 120996-120996 (2021-07-20)
One of the major challenges within the emerging field of injectable stem cell therapies for articular cartilage (AC) repair is the retention of sufficient viable cell numbers at the site of injury. Even when delivered via intra-articular injection, the number
Quantitative molecular sensing in biological tissues: an approach to non-invasive optical characterization
Chandra M, et al.
Optics Express, 14(13), 6157-6171 (2006)
Aging and the skin
Brocklehurst's Textbook of Geriatric Medicine and Gerontology, 133-137 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service