BAF199210001
Bel-Art® Extended Handle Pestle
cone tip polypropylene, pack of 6 pieces
Synonym(s):
grinding and crushing equipment, plastic pestle, tool for manually grinding solid samples
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.74
material
cone tip polypropylene
packaging
pack of 6 pieces
manufacturer/tradename
Bel-Art F19921-0001
L × diam.
21 cm × 4.8 mm
Related Categories
General description
Polypropylene pestle for homogenizing in centrifuge, test and grinding tubes
- Fits conical shaped 1.5 ml tubes
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)