Skip to Content
Merck
All Photos(3)

Key Documents

AV45746

Sigma-Aldrich

Anti-CBS antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Cystathionine-β-synthase, Anti-HIP4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

60 kDa

species reactivity

bovine, horse, rabbit, dog, guinea pig, human, mouse, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CBS(875)

Immunogen

Synthetic peptide directed towards the N terminal region of human CBS

Biochem/physiol Actions

Cystathionine β-synthase (CBS) is an important enzyme in the transsulfuration metabolic pathway. It transforms homocysteine to cystathionine, which is then converted to cysteine, required for production of the chief retinal antioxidant glutathione (GSH).

Sequence

Synthetic peptide located within the following region: RCIIVMPEKMSSEKVDVLRALGAEIVRTPTNARFDSPESHVGVAWRLKNE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phase II study of mitoxantrone in patients with mesothelioma: a National Cancer Institute of Canada Clinical Trials Group Study.
E A Eisenhauer et al.
Cancer treatment reports, 70(8), 1029-1030 (1986-08-01)
Y F Zhou et al.
Journal of dairy science, 101(12), 11384-11395 (2018-10-15)
Insufficient supply of Met and choline (Chol) around parturition could compromise hepatic metabolism and milk protein synthesis in dairy cows. Mechanistic responses associated with supply of Met or Chol in primary liver cells enriched with hepatocytes (PHEP) from cows have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service