Skip to Content
Merck
All Photos(2)

Documents

A3515

Sigma-Aldrich

Adenosine 2′,5′-diphosphate–Agarose

lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56

form

lyophilized powder

Quality Level

extent of labeling

1-5 μmol per mL

matrix

cross-linked 4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

N-6

matrix spacer

8 atoms

storage temp.

−20°C

Related Categories

Application

Adenosine 2′,5′-diphosphate Agarose (2′,5′-ADP agarose) has been used to study NADPH-cytochrome P450 oxidoreductase (CPR), which plays an important role in the insecticide resistance of Anopheles gambiae (a major malaria vector). In particular, Anopheles gambiae CPR (AgCPR) displayed a decreased affinity to adenosine 2′,5′-phosphate when compared to human CPR (hCPR) determined by isothermal titration calorimetry. This may be important for the development of more advanced insecticides that selectively target mosquito CPR.

Physical form

Lyophilized powder stabilized with lactose

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alex Grunau et al.
Biochemistry, 46(28), 8244-8255 (2007-06-22)
A combination of mutagenesis, calorimetry, kinetics, and small-angle X-ray scattering (SAXS) has been used to study the mechanism of ligand binding energy propagation through human cytochrome P450 reductase (CPR). Remarkably, the energetics of 2',5'-ADP binding to R597 at the FAD-binding
Lu-Yun Lian et al.
PloS one, 6(5), e20574-e20574 (2011-06-10)
NADPH-cytochrome P450 oxidoreductase (CPR) plays a central role in chemical detoxification and insecticide resistance in Anopheles gambiae, the major vector for malaria. Anopheles gambiae CPR (AgCPR) was initially expressed in Eschericia coli but failed to bind 2',5'-ADP Sepharose. To investigate
Alex Grunau et al.
Biochemistry, 45(5), 1421-1434 (2006-02-01)
The thermodynamics of coenzyme binding to human cytochrome P450 reductase (CPR) and its isolated FAD-binding domain have been studied by isothermal titration calorimetry. Binding of 2',5'-ADP, NADP(+), and H(4)NADP, an isosteric NADPH analogue, is described in terms of the dissociation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service