Skip to Content
Merck
All Photos(1)

Key Documents

A112000

Sigma-Aldrich

DMT-dA(tac) Phosphoramidite

Synonym(s):

DMT-dA(tac) amidite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H62N7O8P
CAS Number:
Molecular Weight:
944.06
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.51
Pricing and availability is not currently available.

type

for DNA synthesis

Quality Level

product line

Proligo Reagents

Assay

≥98% (31P-NMR)
≥98.0% (reversed phase HPLC)

form

powder

technique(s)

oligo synthesis: suitable

λ

conforms (UV/VIS Identity)

nucleoside profile

base: deoxyadenosine
base protecting group: TAC
2' protecting group: none
5' protecting group: DMT
deprotection: fast

storage temp.

-10 to -25°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H](C[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)n3cnc4c(NC(=O)COc5ccccc5)ncnc34)(c6ccccc6)c7ccc(OC)cc7

InChI

1S/C48H54N7O8P/c1-33(2)55(34(3)4)64(61-27-13-26-49)63-41-28-44(54-32-52-45-46(50-31-51-47(45)54)53-43(56)30-59-40-16-11-8-12-17-40)62-42(41)29-60-48(35-14-9-7-10-15-35,36-18-22-38(57-5)23-19-36)37-20-24-39(58-6)25-21-37/h7-12,14-25,31-34,41-42,44H,13,27-30H2,1-6H3,(H,50,51,53,56)/t41-,42+,44+,64?/m0/s1

InChI key

INUFZOANRLMUCW-SDNJGBRXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DMT-dA(tac) Phosphoramidite is a TAC Protected Phosphoramidite. Substitution of standard protecting groups with the labile TAC (tert.- butylphenoxyacetyl) protecting group results in ultra-fast and easy deprotection under mild conditions, suitable for oligonucleotides with baselabile monomers and reporters as well as in-situ synthesis schemes on glass surfaces.

Key Features of TAC Chemistry are -

  • Deprotection of the TAC group is ultra-fast: complete deprotection in concentrated ammonia occurs within 15 minutes at 55 °C or two hours at room temperature
  • Compatible with the AMA deprotection reagent (a mixture of ≥25% ammonia in water with 40% aqueous methylamine I/I, v/v)
  • Highly soluble in acetonitrile. No need to add co-solvents such as dimethylformamide or methylene chloride
  • Suitable for the synthesis of oligomers with base-labile units e.g., dyes and modifiers, because of less exposure to ammonia and the possibility of room temperature deprotection
  • No change is required in the reagents commonly used for DNA synthesis, except that Proligo′s Fast Deprotection Cap A solution is used instead of Cap A solution •
  • The application of dA(tac) minimizes depurination and improves the quality of oligonucleotides

Features and Benefits

  • The deprotection of oligonucleotide synthesis products with the AMAreagent is ultra-fast: complete deprotection requires 10 minutes at 65 °C
  • Side reactions at C-monomers through transamination are eliminated
  • Not compatible with some base-labile modified nucleosides
  • dC(tac)-phosphoramidite can directly substitute for dC(bz)-phosphoramidite
  • No change is required in the reagents commonly used for DNA synthesis.Acetonitrile is used to dissolve phosphoramidite. The standard aceticanhydride capping reagent can be employed.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service