Skip to Content
Merck
All Photos(1)

Documents

12138C

Sigma-Aldrich

Bovine Calf Serum

Iron-Fortified, USA origin, from formula-fed bovine calves, sterile-filtered, suitable for cell culture

Synonym(s):

Calf Serum, BCS, bovine calf sera, calf sera, calf serum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.71

biological source

bovine (calf) (formula-fed)

Quality Level

sterility

sterile-filtered

composition

hemoglobin, ≤35 mg/dL

origin

USA origin

technique(s)

cell culture | mammalian: suitable

impurities

≤10 EU/mL endotoxin

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Iron-supplemented bovine serum as an alternative to fetal bovine serum in the CHO/HGPRT mutation assay, Oberly, et. al. Development of a more rapid, reduced serum culture system for Caco-2 monolayers and application to the biopharmaceutics classification system. Lentz et. al.

Testing Method

Endotoxin tested

Preparation Note

Each lot is supplemented to a constant iron concentration.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T J Oberly et al.
Mutation research, 244(2), 105-109 (1990-06-01)
Iron-supplemented bovine calf serum (ICS) was found to be a viable alternative to fetal bovine serum (FBS) in the growth promotion and cloning efficiency of Chinese hamster ovary (CHO) cells that are used in the HGPRT mutation assay. Suspension cultures
D R Dahling et al.
Journal of virological methods, 27(3), 287-294 (1990-03-01)
Two studies comparing sewage-isolated and laboratory stock viruses were conducted to determine if alternative forms of serum or serum extenders could be used in place of fetal bovine serum without a significant loss of viral titer. In the first study
K A Lentz et al.
International journal of pharmaceutics, 200(1), 41-51 (2000-06-14)
The objectives were: (1) to develop a more rapid, reduced serum culture system for Caco-2 monolayers, relative to the traditional 21-day, 10% fetal bovine serum (FBS) system; and (2) to determine the biopharmaceutical drug classification of an oral therapeutic agent
Wei Wang et al.
PloS one, 10(4), e0119005-e0119005 (2015-04-24)
Conjugated linoleic acid (CLA) reduces adiposity in human and mouse adipocytes. This outcome is achieved through a variety of biological responses including increased energy expenditure and fatty acid oxidation, increased inflammation, repression of fatty acid biosynthesis, attenuated glucose transport, and

Articles

Find answers to frequently asked questions (FAQs) about FBS and other serum products, and links to related products.

Find answers to frequently asked questions (FAQs) about FBS and other serum products, and links to related products.

Find answers to frequently asked questions (FAQs) about FBS and other serum products, and links to related products.

Find answers to frequently asked questions (FAQs) about FBS and other serum products, and links to related products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service