Key Documents
Z801240
Hellma® aluminum spacer
38mmx12.5mmx8 mm, to fit cells with 2 mm light path into 10 mm cell holder
About This Item
UNSPSC Code:
12000000
NACRES:
SB.32
Pricing and availability is not currently available.
manufacturer/tradename
Hellma 13.102
technique(s)
UV/Vis spectroscopy: suitable
photometry: suitable
Legal Information
Hellma is a registered trademark of Hellma GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)