Skip to Content
Merck
All Photos(1)

Key Documents

Y0000819

Marbofloxacin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

9-Fluoro-2,3-dihydro-3-methyl-10-(4-methyl-piperazino)-7-oxo-7H-pyrido[1,2,3-ij][1,2,4]benzoxadiazine-6-carboxylic acid, Zeniquin®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19FN4O4
CAS Number:
Molecular Weight:
362.36
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

marbofloxacin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CN1CCN(CC1)c2c(F)cc3C(=O)C(=CN4N(C)COc2c34)C(O)=O

InChI

1S/C17H19FN4O4/c1-19-3-5-21(6-4-19)14-12(18)7-10-13-16(14)26-9-20(2)22(13)8-11(15(10)23)17(24)25/h7-8H,3-6,9H2,1-2H3,(H,24,25)

InChI key

BPFYOAJNDMUVBL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Marbofloxacin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Legal Information

Zeniquin is a registered trademark of Pfizer, Inc.

related product

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Vilalta et al.
Journal of veterinary pharmacology and therapeutics, 37(6), 542-549 (2014-06-07)
This study evaluated the theoretical clinical outcome of three marbofloxacin posology regimens in two groups of pigs (weaners and fatteners) for the treatment of Actinobacillus pleuropneumoniae (App) and Haemophilus parasuis (Hp) infection and the appearance of resistant bacteria due to
Xiaoqiang Liu et al.
Veterinary microbiology, 174(3-4), 514-522 (2014-12-04)
This study explored and compared the mechanisms and selective concentration of resistance between a 3rd (pradofloxacin) and 2nd (ciprofloxacin) generation fluoroquinolone. Pradofloxacin- and ciprofloxacin-resistant mutants were selected by stepwise exposure of Escherichia coli (E. coli) to escalating concentrations of pradofloxacin
Seung Ryul Han et al.
Biochemical and biophysical research communications, 448(4), 397-402 (2014-05-06)
Danofloxacin is a synthetic fluoroquinolone with broad spectrum antibacterial activity that is used for the treatment of respiratory diseases in animal husbandry. However, danofloxacin has many adverse reactions and is toxic to humans. Especially, it detrimentally affects muscle, central nerve
Guyue Cheng et al.
Analytical and bioanalytical chemistry, 406(30), 7899-7910 (2014-10-31)
Fluoroquinolones (FQNs) are broad-spectrum antibacterial agents widely used in animal husbandry and aquaculture. The residues and antimicrobial resistance of such antibiotics are a major public health concern. To realize multianalyte detection of FQN residues, a genetically modified bacterium, Escherichia coli
F Yang et al.
Journal of veterinary pharmacology and therapeutics, 37(6), 579-588 (2014-06-07)
Residue depletion of marbofloxacin in broiler chicken after oral administration at 5 mg/kg/day for three consecutive days was studied in this study. The areas under the concentration-time curve from 0 h to ∞ (AUC0-∞ s) of marbofloxacin in tissues and plasma were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service