Skip to Content
Merck
All Photos(1)

Key Documents

Y0000376

Dipivefrine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

[2-(2,2-Dimethylpropanoyloxy)-4-[1-hydroxy-2-(methylamino)ethyl]phenyl] 2,2-dimethylpropanoate hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H29NO5 · HCl
CAS Number:
Molecular Weight:
387.90
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

dipivefrine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C19H29NO5.ClH/c1-18(2,3)16(22)24-14-9-8-12(13(21)11-20-7)10-15(14)25-17(23)19(4,5)6;/h8-10,13,20-21H,11H2,1-7H3;1H

InChI key

VKFAUCPBMAGVRG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dipivefrine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hojung Choi et al.
Journal of veterinary science, 12(1), 99-101 (2011-03-04)
Color Doppler imaging (CDI) was carried out to evaluate the effects of anti-glaucoma drugs on ophthalmic circulation using CDI-derived resistive index (RI) values. CDI was performed on nine Beagle dogs, and RI values were calculated for the medial long posterior
M R Reyes et al.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics, 14(3), 229-237 (1998-07-22)
Previous studies indicate that there may be differences in the protein composition of the aqueous humor in normal and glaucomatous human eyes. These differences in protein composition and concentration may be due to the topical antiglaucoma medications used to treat
F Neu
Bulletin de la Societe belge d'ophtalmologie, (304)(304), 71-76 (2007-08-28)
Cystoid macular edema is a known side effect of different systemic and local medications. Nicotinic acid used as a hypolipemiant agent can cause cystoid macular edema. Local adrenergic antiglaucomatous drugs as well as prostaglandin analogs can induce cystoid macular edema
I Widengård et al.
The British journal of ophthalmology, 82(4), 404-406 (1998-06-26)
To investigate the effect on intraocular pressure (IOP) and aqueous flare of topical applications of latanoprost and dipivefrin alone or combined. 22 patients with open angle glaucoma or ocular hypertension were included in a 4 week open label study. Median
W E Gillies et al.
Australian and New Zealand journal of ophthalmology, 26(3), 215-218 (1998-08-26)
We assessed the long-term additive effect of topical adrenaline or adrenaline precursor and beta-blockers because of doubts as to the presence and persistence of such an effect. In 43 patents (20 males and 23 females; mean (+/- SD) age 68.4+/-11.5

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service