Skip to Content
Merck
All Photos(1)

Key Documents

W384712

Sigma-Aldrich

L-Lysine hydrochloride

natural, FG

Synonym(s):

(2S)-2,6-Diaminohexanoic acid hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N2O2 · HCl
CAS Number:
Molecular Weight:
182.65
EC Number:
MDL number:
UNSPSC Code:
12352209
Flavis number:
17.031
NACRES:
NA.21
Organoleptic:
odorless
grade:
FG
Halal
Kosher
natural
biological source:
corn
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

corn

Quality Level

grade

FG
Halal
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.320

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

odorless

SMILES string

[Cl-].N[C@@H](CCCCN)C(=O)O.[H+]

InChI

1S/C6H14N2O2.ClH/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1H/t5-;/m0./s1

InChI key

BVHLGVCQOALMSV-JEDNCBNOSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suzanne E Dettling et al.
Current protocols in chemical biology, 12(4), e86-e86 (2020-11-17)
Reverse-polarity activity-based protein profiling (RP-ABPP) is a chemical proteomics approach that uses nucleophilic probes amenable to "click" chemistry deployed into living cells in culture to capture, immunoprecipitate, and identify protein-bound electrophiles. RP-ABPP is used to characterize the structure and function
Thomas James O'Brien et al.
F1000Research, 10, 801-801 (2021-09-25)
The airways of people with cystic fibrosis (CF) are often chronically colonised with a diverse array of bacterial and fungal species. However, little is known about the relative partitioning of species between the planktonic and biofilm modes of growth in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service