Skip to Content
Merck
All Photos(1)

Documents

W365820

Sigma-Aldrich

1,4-Cineole

≥95%, FG

Synonym(s):

1-Isopropyl-4-methyl-7-oxabicyclo[2.2.1]heptane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
FEMA Number:
3658
Beilstein:
104974
EC Number:
Council of Europe no.:
11225
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
3.007
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

≥95%

refractive index

n20/D 1.445 (lit.)

bp

65 °C/16 mmHg (lit.)

mp

−46 °C (lit.)

density

0.887 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; camphoraceous; minty; cooling; pine

SMILES string

CC(C)C12CCC(C)(CC1)O2

InChI

1S/C10H18O/c1-8(2)10-6-4-9(3,11-10)5-7-10/h8H,4-7H2,1-3H3/t9-,10+

InChI key

RFFOTVCVTJUTAD-AOOOYVTPSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Solvent-free dehydration, cyclization, and hydrogenation of linalool with a dual heterogeneous catalyst system to generate a high-performance sustainable aviation fuel.: A novel approach utilizing 1,4-Cineole in the synthesis of sustainable aviation fuel through chemical transformations of linalool (Keller et al., 2022).

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service