Skip to Content
Merck
All Photos(1)

Key Documents

PHR3611

Supelco

N-Nitrosomethylphenylamine (NMPA)

certified reference material, pharmaceutical secondary standard

Synonym(s):

N-Methyl-N-nitrosoaniline, N-Methyl-N-nitrosobenzenamine, N-Methyl-N-phenylnitrosamine, NMPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8N2O
CAS Number:
Molecular Weight:
136.15
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1466607

form

methanol solution

CofA

current certificate can be downloaded

packaging

pkg of 1 mL

application(s)

pharmaceutical

storage temp.

-10 to -25°C

SMILES string

N(N=O)(C)c1ccccc1

InChI

1S/C7H8N2O/c1-9(8-10)7-5-3-2-4-6-7/h2-6H,1H3

InChI key

MAXCWSIJKVASQC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

N-Nitrosomethylphenylamine (NMPA) pharmaceutical secondary reference standard is used for impurity analysis of NMPA in pharmaceutical formulations using supercriticalfluid chromatography (SFC).[1]

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Solvent

Description

    1000 ug/mL solution in Methanol

related product

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 1B - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analytical lifecycle management for comprehensive and universal nitrosamine analysis in various pharmaceutical formulations by supercritical fluid chromatography
Schmidtsdorff, S et al.
Journal of Pharmaceutical and Biomedical Analysis, 197, 113960-113960 (2021)

Articles

Discover the efficient separation six nitrosamine impurities with high reproducibility and precise detection to ensure accurate analysis for regulatory compliance.

Discover the efficient separation six nitrosamine impurities with high reproducibility and precise detection to ensure accurate analysis for regulatory compliance.

Discover the efficient separation six nitrosamine impurities with high reproducibility and precise detection to ensure accurate analysis for regulatory compliance.

Discover the efficient separation six nitrosamine impurities with high reproducibility and precise detection to ensure accurate analysis for regulatory compliance.

See All

Related Content

Accurately detect and quantify trace nitrosamines (NDMA, NDEA, NEIPA, NDIPA, NDBA, etc.) in pharmaceutical drugs using our complete product portfolio and application guides for LC-MS and GC-MS. Order high-quality reference standards, columns, filters & more.

Accurately detect and quantify trace nitrosamines (NDMA, NDEA, NEIPA, NDIPA, NDBA, etc.) in pharmaceutical drugs using our complete product portfolio and application guides for LC-MS and GC-MS. Order high-quality reference standards, columns, filters & more.

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Hi, We want to learn the concentration of the liquid reference standard. Could you please share the concentration and solvent of the NMPA reference solution ?

    1 answer
    1. This product is a solution of nominally 1000 µg/mL. The solvent is methanol. The accurate concentration can be found on the lot specific Certificate of Analysis. A Certificate can be accessed in the DOCUMENTATION section under the 'Certificate of Analysis' tab:
      https://www.sigmaaldrich.com/product/sial/phr3611#product-documentation

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service