Skip to Content
Merck
All Photos(1)

Key Documents

PHR1340

Supelco

Pyrilamine maleate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Pyrilamine maleate salt, Mepyramine maleate salt, N-(4-Methoxyphenyl)methyl-N′,N′-dimethyl-N-(2-pyridinyl)-1,2-ethanediamine maleate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H23N3O · C4H4O4
CAS Number:
Molecular Weight:
401.46
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. M0500000
traceable to USP 1588004

API family

mepyramine, pyrilamine

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

OC(=O)\C=C/C(O)=O.COc1ccc(CN(CCN(C)C)c2ccccn2)cc1

InChI

1S/C17H23N3O.C4H4O4/c1-19(2)12-13-20(17-6-4-5-11-18-17)14-15-7-9-16(21-3)10-8-15;5-3(6)1-2-4(7)8/h4-11H,12-14H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

JXYWFNAQESKDNC-BTJKTKAUSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Pyrilamine maleate is a first generation antihistamine drug, widely used to reduce all symptoms of cold and is also employed to reduce allergies both parenterally and locally.[1]

Application

Pyrilamine maleate may be used pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using high-performance liquid chromatography technique[2] and liquid chromatography technique.[1]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA1092 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of acetaminophen, pamabrom and pyrilamine maleate in pharmaceutical formulations using stability indicating HPLC assay method
Saleem A, et al.
Journal of the Mexican Chemical Society, 59(2), 93-98 (2015)
Simultaneous determination of dextromethorphan hydrobromide, pyrilamine maleate and sodium benzoate in a cough cold syrup by LC
Louhaichi RM, et al.
Chromatographia, 69(5), 421-427 (2009)
Trace analysis of the antihistamines methapyrilene hydrochloride, pyrilamine maleate and triprolidine hydrochloride monohydrate in animal feed, human urine and wastewater by high-performance liquid chromatography and gas chromatography with nitrogen?phosphorus detection.
Thompson C H and Holder L C.
Journal of Chromatography A, 283, 251-264 (1984)
Validation of a HPLC quantification of acetaminophen, phenylephrine and chlorpheniramine in pharmaceutical formulations: capsules and sachets.
Marin A, et al.
Journal of Pharmaceutical and Biomedical Analysis, 29(4), 701-714 (2002)
Lingli Yang et al.
The Journal of investigative dermatology, 134(8), 2105-2113 (2014-03-01)
Histamine is thought to have a critical role in the synthesis of extracellular matrix in skin and may be involved in tissue remodeling of allergic diseases. Recent studies revealed that periostin, a matricelluar protein, contributed to tissue remodeling; however, a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service