NIST635A
Portland Cement (Blended with Slag)
NIST®SRM®
About This Item
Recommended Products
grade
certified reference material
packaging
pkg of 25 g
manufacturer/tradename
NIST®
format
matrix material
storage temp.
15-25°C
General description
Other Notes
Silicon Dioxide (SiO2), Aluminum Trioxide (Al2O3), Iron (III) Oxide (Fe2O3), Calcium Oxide (CaO), Magnesium Oxide (MgO), Sulfur Trioxide (SO3), Sodium Oxide (Na2O), Potassium Oxide (K2O), Titanium Dioxide (TiO2), Phosphorus Pentoxide (P2O5), Manganese Trioxide (Mn2O3), Chromium Trioxide (Cr2O3), Zinc Oxide (ZnO), Strontium Oxide (SrO), Barium Oxide (BaO), Chlorine (Cl)
Reference/Informational value is provided for the following analytes.
Insoluble residue, Calcium oxide (CaO) , Sulfide Sulfur (S²?), Fluorine (F)
See certificate for values and more details at nist.gov/SRM.
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
EU REACH Annex XVII (Restriction List)
EU REACH Annex XIV (Authorisation List)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service