Skip to Content
Merck
All Photos(2)

Documents

91443

Supelco

Glycyrrhizic acid ammonium salt

analytical standard, suitable for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H62O16 · NH3
CAS Number:
Molecular Weight:
839.96
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

N.CC1(C)[C@H](CC[C@@]2(C)[C@H]1CC[C@]3(C)[C@@H]2C(=O)C=C4[C@@H]5C[C@](C)(CC[C@]5(C)CC[C@@]34C)C(O)=O)O[C@H]6O[C@@H]([C@@H](O)[C@H](O)[C@H]6O[C@@H]7O[C@@H]([C@@H](O)[C@H](O)[C@H]7O)C(O)=O)C(O)=O

InChI

1S/C42H62O16.H3N/c1-37(2)21-8-11-42(7)31(20(43)16-18-19-17-39(4,36(53)54)13-12-38(19,3)14-15-41(18,42)6)40(21,5)10-9-22(37)55-35-30(26(47)25(46)29(57-35)33(51)52)58-34-27(48)23(44)24(45)28(56-34)32(49)50;/h16,19,21-31,34-35,44-48H,8-15,17H2,1-7H3,(H,49,50)(H,51,52)(H,53,54);1H3/t19-,21-,22-,23-,24-,25-,26-,27+,28-,29-,30+,31+,34-,35-,38+,39-,40-,41+,42+;/m0./s1

InChI key

ILRKKHJEINIICQ-OOFFSTKBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Glycyrrhizic acid ammonium salt may be used as a primary chemical reference standard for the quantitative determination of glycyrrhizic acid in Glycyrrhiza uralensis fisch. ex DC. plant root using high performance liquid chromatography(HPLC).
Glycyrrhizic acid ammonium salt may be used as an analytical standard for the determination of glycyrrhizic acid in biological fluids using micellar electrokinetic chromatographic technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: glycyrrhiza

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of glycyrrhizic acid and 18-β-glycyrrhetinic acid in biological fluids by micellar electrokinetic chromatography
Wang P, et al.
Journal of Chromatography A, 811(1-2), 219-224 (1998)
The determination of glycyrrhizic acid in Glycyrrhiza uralensis Fisch. ex DC.(Zhi Gan Cao) root and the dried aqueous extract by LC?DAD
Hennell.R.J, et al.
Journal of Pharmaceutical and Biomedical Analysis, 47, 494-500 (2008)
Yu Okuma et al.
Neuropharmacology, 85, 18-26 (2014-05-27)
Glycyrrhizin (GL) is a major constituent of licorice root and has been suggested to inhibit the release of high mobility group box-1 (HMGB1), a protein considered representative of damage-associated molecular patterns. We found that GL bound HMGB1 but not RAGE
Yusuke Izutani et al.
Carbohydrate research, 392, 25-30 (2014-05-23)
The structural properties of glycyrrhizic acid, a sweet-tasting constituent of Glycyrrhiza glabra, and its interaction with cyclodextrins were analyzed using dynamic light scattering, isothermal titration calorimetry, and NMR. The dynamic light scattering and NMR studies showed that glycyrrhizic acid forms
Hsin-Yi Lo et al.
PloS one, 9(8), e104650-e104650 (2014-08-19)
Diabetes is a chronic metabolic disorder that has a significant impact on the health care system. The reduction of glycated hemoglobin A1c is highly associated with the improvements of glycemic control and diabetic complications. In this study, we identified a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service