Skip to Content
Merck
All Photos(1)

Key Documents

91286

Supelco

Multi Cation Standard 1 for IC

TraceCERT®, in 0.1% nitric acid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

composition

Ca, 1000 mg/L
K, 200 mg/L
Li, 50 mg/L
Mg, 200 mg/L
Na, 200 mg/L
NH4, 400 mg/L

concentration

in 0.1% nitric acid

technique(s)

ion chromatography: suitable

application(s)

environmental
food and beverages

format

multi-component solution

Application


  • Natural killer cytotoxicity in myalgic encephalomyelitis/chronic fatigue syndrome (ME/CFS): a multi-site clinical assessment of ME/CFS (MCAM) sub-study: This study highlights the application of Multi Cation Standard 1 for IC in clinical research, specifically in the analysis of immune system functionality in ME/CFS patients. It showcases how ion chromatography calibration standards are crucial for precise and reliable measurements in biomedical research (Querec TD, et al., 2023).

  • Sub-1 mL sample requirement for simultaneous determination of 17 organic and inorganic anions and cations in Antarctic ice core samples by dual capillary ion chromatography: This publication illustrates the use of Multi Cation Standard 1 for IC in environmental science, emphasizing its role in the simultaneous analysis of multiple ions in extremely small volume samples from Antarctic ice cores. This study underlines the versatility of IC multi-cation calibration standards in various scientific fields, from environmental to pharmaceutical QC (Sanz Rodriguez E, et al., 2019).

Linkage

Visit the IC Portal to learn more

Analysis Note

For the preparation of this CRM only materials of highest purity were used.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service