Skip to Content
Merck
All Photos(1)

Key Documents

89630

Supelco

p-Toluidine

for spectrophotometric det. of Au, Tl(III), W, ≥99.0%

Synonym(s):

4-Aminotoluene, 4-Methylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4NH2
CAS Number:
Molecular Weight:
107.15
Beilstein:
471281
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21
technique(s):
UV/Vis spectroscopy: suitable
bp:
200 °C (lit.)
vapor pressure:
1 hPa ( 42 °C)
7 hPa ( 68 °C)

vapor density

3.9 (vs air)

Quality Level

vapor pressure

1 hPa ( 42 °C)
7 hPa ( 68 °C)

Assay

≥99.0% (GC)
≥99.0%

form

crystals

autoignition temp.

899 °F

quality

for spectrophotometric det. of Au, Tl(III), W

expl. lim.

6.6 %

technique(s)

UV/Vis spectroscopy: suitable

bp

200 °C (lit.)

mp

41-46 °C (lit.)
42-45 °C

solubility

ethanol: 1 g/10 mL (Hot)

density

0.973 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(N)cc1

InChI

1S/C7H9N/c1-6-2-4-7(8)5-3-6/h2-5H,8H2,1H3

InChI key

RZXMPPFPUUCRFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for the spectrophotometric det. of Tl(III), Au and W

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Skin Sens. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G. Pfrepper
Fresenius Journal of Analytical Chemistry, 193, 179-179 (1963)
N. Mishra et al.
Analyst, 112, 1131-1131 (1987)
K.S. Patel et al.
Analytical Chemistry, 58, 1547-1547 (1986)
Paul G Stevenson et al.
Journal of chromatography. A, 1218(45), 8255-8263 (2011-10-11)
Several simple techniques are presented for the identification of the boundaries of chromatographic peaks. These methods provide a significant reduction in the time needed to perform the rapid, automatic calculation of the central peak moments and to evaluate the quality
Shunji Ito et al.
The Journal of organic chemistry, 70(6), 2285-2293 (2005-03-12)
[reaction: see text] N,N-Di(6-azulenyl)-p-toluidine (1a) and N,N,N',N'-tetra(6-azulenyl)-p-phenylenediamine (2a) and their derivatives with 1,3-bis(ethoxycarbonyl) substituents on each 6-azulenyl group (1b and 2b) were prepared by Pd-catalyzed amine azulenylation and characterized as a study into new aromatic amines for multistage amphoteric redox

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service