Skip to Content
Merck
All Photos(1)

Key Documents

87115

Supelco

Methyl tetracosanoate

analytical standard

Synonym(s):

Lignoceric acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)22COOCH3
CAS Number:
Molecular Weight:
382.66
Beilstein:
1797895
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

232 °C/4 mmHg (lit.)

mp

58-60 °C (lit.)

format

neat

functional group

ester

shipped in

ambient

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCC(=O)OC

InChI

1S/C25H50O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25(26)27-2/h3-24H2,1-2H3

InChI key

XUDJZDNUVZHSKZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl tetracosanoate may be used as an external standard to study the pentafluorobenzylation reaction of estradiol in a biphasic methylene chloride/sodium hydroxide system and in the absence of phase transfer agents by gas chromatography technique (GC).
Methyl tetracosanoate may be used as an analytical reference standard for the determination of the analyte in biodiesel produced from yellow horn oil using high-performance liquid chromatography technique.[1]
Methyl tetracosanoate may be used as an external standard to study the pentafluorobenzylation reaction of estradiol in a biphasic methylene chloride/sodium hydroxide system and in the absence of phase transfer agents by gas chromatography technique(GC).[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hawley's Condensed Chemical Dictionary, (16) (2016)
Specific pentafluorobenzylation of phenols in a biphasic system
Rosenfeld.M.J and Crocco.L.J
Analytical Chemistry, 50, 701-704 (1978)
Kusampudi Shilpa et al.
Biotechnology letters, 31(12), 1837-1841 (2009-08-21)
A methanolic extract of Costus pictus (CPME) showed optimum anti-diabetic activity at 100 ng/ml. Bioactivity-guided purification of CPME led to the isolation of methyl tetracosanoate (MT) which showed an optimum glucose uptake at 1 ng/ml. CPME at 10 mug/ml inhibited
Determination of fatty acid methyl esters in biodiesel produced from yellow horn oil by LC
Fu J-Y, et al.
Chromatographia, 67(1-2), 9-14 (2008)
M A Lambert et al.
Journal of clinical microbiology, 23(4), 731-736 (1986-04-01)
After growth and experimental conditions were established, the mycolic acid cleavage products, constituent fatty acids, and alcohols of representative strains of Mycobacterium tuberculosis, M. smegmatis, M. fortuitum complex, M. kansasii, M. gordonae, and M. avium complex were determined by capillary

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service