Skip to Content
Merck
All Photos(1)

Key Documents

70152

Millipore

Plate Count Agar

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Casein-peptone Dextrose Yeast Agar, Standard Methods Agar, Tryptone Glucose Yeast Extract Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171605
NACRES:
NA.74

Agency

according to GB 4789.2-2016
according to ISO 17410:2019
according to ISO 4833-1:2013
according to ISO 4833-2:2013

Quality Level

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 9.0 g/L
dextrose, 1.0 g/L
tryptone, 5.0 g/L
yeast extract, 2.5 g/L

manufacturer/tradename

NutriSelect® Plus

packaging

pkg of -2.5 kg
pkg of -500 g

storage condition

(Dry place, tightly sealed containers)

technique(s)

microbiological culture: suitable

final pH

7.0±0.2 (25 °C)

application(s)

agriculture
bioburden testing
environmental
food and beverages
water monitoring

microbiology

storage temp.

2-25°C

suitability

nonselective for Bacillus spp.
nonselective for Escherichia coli
nonselective for Lactobacillus spp.
nonselective for Lactococcus spp.
nonselective for Listeria spp.
nonselective for Staphylococcus spp.
nonselective for Streptococcus spp.
nonselective for bacteria (General Media)
nonselective for coliforms

General description

Plate Count Agar is the recommended medium forthe isolation of microorganisms in milk and other dairy products. The enzymaticdigest of casein (tryptone) with a high level of free amino acids is a nitrogensource. Yeast extract primarily supplies the B-complex vitamins. Glucose is anenergy source for the growth of bacteria, while agar is the solidifying agent.Plate Count Agar is also suitable for determining bacterial count from sterilerooms.

Application

Plate Count Agar is a non-selective medium for the plate count of microorganisms in water, food, milk, dairy products, beverages, animal feed, and other materials.

Preparation Note

Suspend 17.5 g in 1 litre of distilled water. Dissolve by bringing to the boil with frequent stirring, mix and distribute into final containers. Sterilize by autoclaving at 121°C for 15 minutes.

Other Notes

Comparison of β-glucuronidase and indole-based direct plating methods for enumeration of unstressed E. coli

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mustafa Durmuş et al.
Journal of food science and technology, 57(3), 895-904 (2020-03-04)
This study was conducted to investigate the effects of nanoemulsions prepared with various concentrations of olive oil on the sensory, chemical and microbiological quality of rainbow trout fillets. The results showed that the shelf life of rainbow trout fillets were
E.W. Frampton et al.
Journal of Food Protection, 53, 933-933 (1990)
Zhongke Sun et al.
3 Biotech, 7(5), 335-335 (2017-09-29)
At present, artificial pit mud (APM) is widely used in Chinese liquor-making industry and plays a particular role in the production of Chinese strong flavor liquor (CSFL). However, APM frequently ages during fermentation, thus becoming unsuitable for sustainable use due

Articles

Some Bacillus species are crucial for advancing our understanding of bacteria, while others can be harmful, acting as spoilage organisms or pathogens. Nevertheless, all these species are significant in biotechnological research.

Some Bacillus species are crucial for advancing our understanding of bacteria, while others can be harmful, acting as spoilage organisms or pathogens. Nevertheless, all these species are significant in biotechnological research.

Microbiological assessment of bacteria found in spices, a potent source for food spoilage and pathogens.

Some Bacillus species are crucial for advancing our understanding of bacteria, while others can be harmful, acting as spoilage organisms or pathogens. Nevertheless, all these species are significant in biotechnological research.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service