Skip to Content
Merck
All Photos(1)

Key Documents

67886

Millipore

Pyrase Test Strips

suitable for clinical testing, food and beverages, microbiology and specific enzyme detection

Synonym(s):

Pyrrolidonyl Peptidase Strips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

Product Name

Pyrase Strips, suitable for microbiology

product line

BioChemika

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

microbe id | specific enzyme detection: suitable

application(s)

clinical testing
food and beverages

microbiology

storage temp.

2-8°C

suitability

Gemella spp.
Enterococcus spp.
Lactococcus spp.
Streptococcus spp.
microbe id | in situ hybridization spp.

General description

Pyrase Strips are for enzymatic test for the detection of pyrrolidonyl peptidase (pyrase). Pyrase is an enzyme produced by all common species of Enterococci and by 98% of Streptococcus pyogenes strains. Detection of pyrase is a sensitive, reliable and rapid method of wide application. It is substantially more efficient than the bacitracine disc tests (Cat. No. 08382) used for the differentiation of S. pyogenes strains and the tests used for grouping of Enterococci. Strip can also be used for the identification of Gram-positive genera, catalase negative cocci, and coccobacilli.
The result of the reaction is read after 2 min.

Application

Pyrase Strips are used for differentiation of Enterococci from the group D Streptococci and differentiation of Streptococcus pyogenes from other haemolytic Streptococci.

Packaging

50 test strips, 1.0 mL reagent

Components

The kit contains plastic strips with a paper zone saturated with chromogenic substrate for the detection of pyrrolidonyl peptidase and a bottle containing a developing reagent.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - STOT SE 2

Target Organs

Immune system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Rapid identification of enterococci by pyrrolidonyl aminopeptidase activity (PYRase).
Mitchell MJ et al.
Diagnostic Microbiology and Infectious Disease, 6, 283-283 (1987)

Articles

For microbiologists the most fundamental stain was developed in 1884 by the Danish bacteriologist Hans Christian Gram.

For microbiologists the most fundamental stain was developed in 1884 by the Danish bacteriologist Hans Christian Gram.

For microbiologists the most fundamental stain was developed in 1884 by the Danish bacteriologist Hans Christian Gram.

For microbiologists the most fundamental stain was developed in 1884 by the Danish bacteriologist Hans Christian Gram.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service