These are the temperature coefficient of variation tables for the five materials:
•Ref. 06949-500ML | Conductivity standard, conductivity 1000 μS/cm at 25 °C
•Ref. 67233-500ML | Conductivity standard, conductivity 10000 μS/cm at 25 °C
•Ref. 56255-500ML | Conductivity standard, conductivity 50000 μS/cm at 25 °C
•Ref. 59777-500ML | Conductivity standard, conductivity 100000 μS/cm at 25 °C
•Ref. 03729-500ML | Conductivity standard, conductivity 200000 μS/cm at 25 °C
56255
Conductivity Standard
conductivity 50000 μS/cm at 25 °C
Synonym(s):
Calibration standard for conductivity meter
About This Item
Recommended Products
grade
analytical standard
Quality Level
shelf life
limited shelf life, expiry date on the label
concentration
2.5-10% KCl (T)
conductivity
49500-50500 μS/cm at 25 °C
density
1.014-1.063 g/cm3 at 20 °C
application(s)
cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care
format
single component solution
Looking for similar products? Visit Product Comparison Guide
Application
standard
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - STOT RE 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
EU REACH Annex XVII (Restriction List)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Do these products have conductivity correction tables at different temperatures?
1 answer-
Helpful?
-
-
Hi, I wonder if u maybe have some kind of table of conductivity corrections at various temperatures? Thanks in advance!
1 answer-
Unfortunately, there is no table for conductivity at different temperatures available. The only value of conductivity shared is on the lot specific Certificate of Analysis where the Conductivity range is determined at 25 C between 49500 - 50500 µS/cm. The exact value will be listed on the Certificate of Analysis.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service