Skip to Content
Merck
All Photos(2)

Key Documents

45633

Supelco

Profluralin

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16F3N3O4
CAS Number:
Molecular Weight:
347.29
Beilstein:
2179006
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCCN(CC1CC1)c2c(cc(cc2[N+]([O-])=O)C(F)(F)F)[N+]([O-])=O

InChI

1S/C14H16F3N3O4/c1-2-5-18(8-9-3-4-9)13-11(19(21)22)6-10(14(15,16)17)7-12(13)20(23)24/h6-7,9H,2-5,8H2,1H3

InChI key

ITVQAKZNYJEWKS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R C Honeycutt et al.
Journal of environmental pathology and toxicology, 3(5-6), 21-33 (1980-06-01)
Recent developments in the technology of removal and characterization of bound residues of two herbicides, nitrofen and profluralin, from plants and soil will be reviewed. 14C-Nitrofen was found to be metabolized into starch in wheat grain as well as bound
A R Jacobson et al.
Chemical research in toxicology, 1(5), 304-311 (1988-09-01)
Fluorine NMR spectroscopy has been used to identify and quantitate metabolites of profluralin appearing in the urine of rats postingestion of the herbicide. The chemical shift of the trifluoromethyl group was shown to be a useful indicator of the chemical
N D Camper et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 15(5), 457-473 (1980-01-01)
The degradation of profluralin [N-(cyclopropylmethyl)-alpha,alpha,alpha-trifluoro-2,6-dinitro-N-propyl-p-toluidine] and trifluralin (alpha,alpha,alpha-trifluoro-2,6-dinitro-N,N-dipropyl-p-toluidine) was studied under aerobic and anaerobic soil conditions. Three soils (Goldsboro loamy sand, Cecil loamy sand, Drummer clay loam) were each treated with 1 ppmw herbicide; anaerobic conditions were maintained by flooding.
M J Arrowood et al.
FEMS microbiology letters, 136(3), 245-249 (1996-03-01)
Despite the evaluation of over 100 antimicrobial drugs, the diarrheal disease cryptosporidiosis has remained refractory to treatment. We report the evaluation of five dinitroaniline herbicides including trifluralin, profluralin, nitralin, pendimethalin, and fluchloralin for anticryptosporidial activity in an in vitro cultivation
T R Edgerton et al.
Journal of analytical toxicology, 9(1), 15-19 (1985-01-01)
A method is presented for the analysis of trace amounts of dinitroaniline herbicides in tissue and excreta. The method employs extraction of the tissue or excreta with organic solvent, clean up by liquid/liquid partitioning or silica gel chromatography, and ultimate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service