Skip to Content
Merck
All Photos(2)

Key Documents

45437

Supelco

Dichlorprop-methyl ester

PESTANAL®, analytical standard

Synonym(s):

Methyl 2-(2,4-dichlorophenoxy)propionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10Cl2O3
CAS Number:
Molecular Weight:
249.09
Beilstein:
6059885
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
NMR: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COC(=O)C(C)Oc1ccc(Cl)cc1Cl

InChI

1S/C10H10Cl2O3/c1-6(10(13)14-2)15-9-4-3-7(11)5-8(9)12/h3-6H,1-2H3

InChI key

SCHCPDWDIOTCMJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yue-Zhong Wen et al.
Journal of environmental sciences (China), 17(2), 237-240 (2005-11-22)
The effect of beta-cyclodextrins (beta-CDs) on the enzymatical hydrolysis of chiral dichlorprop methyl ester (DCPPM) was studied. Four kinds of beta-cyclodextrins (beta-cyclodextrin, Partly methylated-CD (PM-beta-CD), hydroxypropyl-cyclodextrin (HP-beta-CD) and carboxymethyl-cyclodextrin (CM-beta-CD)) were used. Compared with 100% DCPPM in the absence of
F G Sánchez et al.
Analytical biochemistry, 239(1), 2-7 (1996-07-15)
This paper describes a fluorescence enzyme-linked immunosorbent assay (ELISA) for the quantification of (+/-)-2-(2, 4-dichlorophen-oxy)propionic methyl ester (dichlorprop methyl ester). Antibodies for dichlorprop methyl ester were produced by immunizing rabbits with a conjugate of dichlorprop methyl ester with bovine serum
Yun Ma et al.
Chirality, 21(4), 480-483 (2008-07-26)
Chiral pesticides currently constitute about 50% of all pesticides dosage used in China, and this ratio is increasing as more complex structures are introduced. Dichlorprop methyl (DCPPM) is a chiral herbicide consisting of a pair of enantiomers. In this study
Yun Ma et al.
Huan jing ke xue= Huanjing kexue, 26(4), 152-155 (2005-10-11)
Enantioselective Degradation of 2,4-Dichlorprop methyl ester was studied by sediment bacteria. One DCPPM-degrading bacteria stain was isolated from contaminated sediments. The EF values were determined by chiral GC. The enantioselectivity of DCPPM biodegradation in aquatic phases were evaluated. Incubation with
Yuezhong Wen et al.
Journal of agricultural and food chemistry, 59(5), 1924-1930 (2011-02-15)
Up to 25% of the current pesticides are chiral, the molecules have chiral centers, but most of them are used as racemates. In most cases, enantiomers of chiral pesticides have different fates in the environment. Knowledge of the function of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service