Skip to Content
Merck
All Photos(1)

Key Documents

41883

Sigma-Aldrich

Rebaudioside N

≥95.0% (HPLC)

Synonym(s):

Kaur-16-en-18-oic acid, 13-[(O-β-D-glucopyranosyl-(1→2)-O-[b-D-glucopyranosyl-(1→3)]-β-D-glucopyranosyl)oxy]-, O-6-deoxy-α-L-mannopyranosyl-(1→2)-O-[β-D-glucopyranosyl-(1→3)]-β-D-glucopyranosyl ester, (4α)-

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H90O32
CAS Number:
Molecular Weight:
1275.29
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.28

Quality Level

Assay

≥95.0% (HPLC)

form

solid

color

white to off-white

application(s)

food and beverages

storage temp.

2-8°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O[C@@H]%10O[C@@H]([C@H]([C@@H]([C@H]%10O)O)O)CO)O[C@@H]9O[C@@H]([C@H]([C@@H]([C@H]9O)O)O)CO)O[C@]32C[C@]4([C@H]([C@]5([C@@H]([C@](CCC5)(C)C(=O)O[C@@H]6O[C@@H]([C@H]([C@@H]([C@H]6O[C@@H]8O[C@H]([C@@H]([C@H]([C@H]8O)O)

InChI

1S/C56H90O32/c1-19-12-55-10-6-26-53(3,8-5-9-54(26,4)52(76)87-50-44(85-46-38(72)34(68)28(62)20(2)77-46)42(32(66)24(16-60)81-50)83-47-39(73)35(69)29(63)21(13-57)78-47)27(55)7-11-56(19,18-55)88-51-45(86-49-41(75)37(71)31(65)23(15-59)80-49)43(33(67)25(17-61)8

InChI key

AKEKAGBWNXIWSS-ZFXKUSSPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Rebaudioside N is a diterpene glycoside isolated from Stevia rebaudiana..

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of Novel Steviol Glycosides from Leaves of Stevia rebaudiana Morita
Ohta, M., et al.
Journal of Applied Glycoscience, 57, 199-209 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service