Skip to Content
Merck
All Photos(2)

Key Documents

34229

Supelco

(Z)-Metominostrobin

PESTANAL®, analytical standard

Synonym(s):

(Z)-α-Methoxyimino-N-methyl-2-phenoxyphenylacetamide, (Z)-2-Methoxyimino-N-methyl-2-(2-phenoxyphenyl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16N2O3
CAS Number:
Molecular Weight:
284.31
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CNC(=O)C(=N/OC)\c1ccccc1Oc2ccccc2

InChI

1S/C16H16N2O3/c1-17-16(19)15(18-20-2)13-10-6-7-11-14(13)21-12-8-4-3-5-9-12/h3-11H,1-2H3,(H,17,19)/b18-15-

InChI key

HIIRDDUVRXCDBN-SDXDJHTJSA-N

General description

Metominostrobin belongs to the class of strobilurins and acts by inhibiting the mitochondrial respiration of the fungus.

Application

(Z)-Metominostrobin may be used as an analytical reference standard for the determination of the analyte in food matrices, agricultural products and water samples by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A simple and sensitive method for simultaneous analysis of multiresidual fungicides in water samples by HPLC-ESI-MS/MS
Wang Y, et al.
Analytical Methods : Advancing Methods and Applications, 4(12), 4146-4154 (2012)
Method development and validation for strobilurin fungicides in baby foods by solid-phase microextraction gas chromatography-mass spectrometry.
Vinas P, et al.
Journal of Chromatography A, 1216(1), 140-146 (2009)
Rapid method for the determination of 180 pesticide residues in foods by gas chromatography/mass spectrometry and flame photometric detection.
Okihashi M, et al.
Journal of Pesticide Science, 30(4), 368-377 (2005)
Validation of multiresidue screening methods for the determination of 186 pesticides in 11 agricultural products using gas chromatography (GC).
Hirahara Y, et al.
Journal of Health Science, 51(5), 617-627 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service