Skip to Content
Merck
All Photos(2)

Key Documents

34169

Supelco

Dibutyl phthalate-3,4,5,6-d4

PESTANAL®, analytical standard

Synonym(s):

Di-n-butyl phthalate-d4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D4-1,2-[CO2(CH2)3CH3]2
CAS Number:
Molecular Weight:
282.37
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.492 (lit.)

bp

340 °C (lit.)

mp

−35 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

mass shift

M+4

SMILES string

[2H]c1c([2H])c([2H])c(C(=O)OCCCC)c(c1[2H])C(=O)OCCCC

InChI

1S/C16H22O4/c1-3-5-11-19-15(17)13-9-7-8-10-14(13)16(18)20-12-6-4-2/h7-10H,3-6,11-12H2,1-2H3/i7D,8D,9D,10D

InChI key

DOIRQSBPFJWKBE-ULDPCNCHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dibutyl phthalate-3,4,5,6-d4 may be used as a standard solution in the determination of phthalic acid esters in soil, using gas chromatography coupled with mass spectrometry (GC/MS).

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soil contamination by phthalate esters in Chinese intensive vegetable production systems with different modes of use of plastic film
Wang J, et al.
Environmental Pollution (Barking, Essex : 1987), 180, 265-262 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service