Skip to Content
Merck
All Photos(1)

Key Documents

32426

Supelco

Cefoperazone dihydrate

VETRANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H27N9O8S2 · 2H2O
CAS Number:
Molecular Weight:
681.70
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O.O.CCN1CCN(C(=O)N[C@@H](C(=O)N[C@H]2[C@H]3SCC(CSc4nnnn4C)=C(N3C2=O)C(O)=O)c5ccc(O)cc5)C(=O)C1=O

InChI

1S/C25H27N9O8S2.2H2O/c1-3-32-8-9-33(21(39)20(32)38)24(42)27-15(12-4-6-14(35)7-5-12)18(36)26-16-19(37)34-17(23(40)41)13(10-43-22(16)34)11-44-25-28-29-30-31(25)2;;/h4-7,15-16,22,35H,3,8-11H2,1-2H3,(H,26,36)(H,27,42)(H,40,41);2*1H2/t15-,16-,22-;;/m1../s1

InChI key

OMYIZCZQILAOGB-BYLXUVCXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cefoperazone dihydrate may be used as an analytical standard for the determination of cefoperazone in raw bovine milk by high-performance liquid chromatographic (HPLC) method based on solid-phase extraction.
Cefoperazone dihydrate may also be used as an analytical standard for the determination of ticarcillin in human biological samples by reversed-phase liquid chromatography method.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of ticarcillin in human plasma and in urine by reversed-phase LC.
La Follette G, et al.
Journal of Pharmaceutical and Biomedical Analysis, 13(2), 159-164 (1995)
Determination of cephalosporins in raw bovine milk by high-performance liquid chromatography.
S?rensen LK and Snor LK
Journal of Chromatography A, 882(1-2), 145-151 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service