Skip to Content
Merck
All Photos(2)

Key Documents

318744

Sigma-Aldrich

Bromophenol Blue solution

0.04 wt. % in H2O

Synonym(s):

3′,3′′,5′,5′′-Tetrabromophenolsulfonephthalein, Bromophenol Blue sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H9Br4NaO5S
CAS Number:
Molecular Weight:
691.94
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

liquid

Quality Level

concentration

0.04 wt. % in H2O

technique(s)

titration: suitable

pH

3.0-4.6, yellow to blue

density

0.985 g/mL at 25 °C

λmax

589 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].Oc1c(Br)cc(cc1Br)C(=C2/C=C(Br)C(=O)C(Br)=C2)\c3ccccc3S([O-])(=O)=O

InChI

1S/C19H10Br4O5S.Na/c20-12-5-9(6-13(21)18(12)24)17(10-7-14(22)19(25)15(23)8-10)11-3-1-2-4-16(11)29(26,27)28;/h1-8,24H,(H,26,27,28);/q;+1/p-1

InChI key

TWKDJMRHSLWRFI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Bromophenol Blue is a tracking dye for nucleic acid or protein electrophoresis in agarose or polyacrylamide gels.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Rapid transfer of DNA from agarose gels to nylon membranes.
Reed KC and Mann DA.
Nucleic Acids Research, 13, 7207-7221 (1985)
Detection of erythrocyte membrane proteins, sialoglycoproteins, and lipids in the same polyacrylamide gel using a double-staining technique.
Dzandu JK, et.al.
Proceedings of the National Academy of Sciences of the USA, 81, 1733-1737 (1984)
Detection of erythrocyte membrane proteins, sialoglycoproteins, and lipids in the same polyacrylamide gel using a double-staining technique.
Dzandu JK, et al.
Proceedings of the National Academy of Sciences of the USA, 81, 1733-1737 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service